Skip to Content
Merck

Skip To

373260

Heat Shock Protein Inhibitor I

Heat Shock Protein Inhibitor I, CAS 218924-25-5 blocks the induction of HSP70, HSP72, and HSP105. Blocks the development of thermotolerance in a dose-dependent manner.

Synonym(s):

Heat Shock Protein Inhibitor I, KNK437, N-Formyl-3,4-methylenedioxy-benzylidine-γ-butyrolactam, Hsp70 Induction Inhibitor I

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C13H11NO4
CAS Number:
Molecular Weight:
245.23
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77
Assay:
≥95% (HPLC)
Form:
powder
Storage condition:
OK to freeze, protect from light


Quality Segment

assay

≥95% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

white to light yellow

solubility

DMSO: 3 mg/mL, olive oil: soluble

shipped in

ambient

storage temp.

−20°C

SMILES string

N1(CCC(=Cc2cc3c(cc2)OCO3)C1=O)C=O

InChI key

LZGGUFLRKIMBDQ-UHFFFAOYSA-N

General description

A benzylidene lactam compound that blocks the induction of heat shock proteins HSP70, HSP72, and HSP105. Blocks the development of thermotolerance in a dose-dependent manner. Does not affect the thermosensitivity of nontolerant cells. Reported to be less toxic and more effective than Quercetin (Cat. No. 551600); and unlike Quercetin, it does not have an affect on the activities of PKA, PKC, or PTK.
A benzylidine lactam compound that inhibits the induction of heat shock proteins, including HSP70, HSP72, and HSP105. Blocks the development of thermotolerance in a dose-dependent manner, but does not affect the thermosensitivity of nontolerant cells. Reported to be less toxic and more effective than Quercetin (Cat. No. 551600), and, unlike quercetin, it does not affect the activities of PKA, PKC, and PTK.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Hsp70
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze at -20°C. Stock solutions are stable for up to 6 weeks at -20°C.

Other Notes

Koishi, M., et al. 2001. Clin. Cancer Res.7, 215.
Yokota, S., et al. 2000. Cancer Res.60, 2942.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Harmful (C)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
385570539160567731
form

powder

form

solid

form

solid

form

solid

assay

≥95% (HPLC)

assay

≥98% (HPLC)

assay

≥90% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

100

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

storage condition

OK to freeze, protect from light

storage condition

OK to freeze, protect from light

storage condition

OK to freeze

storage condition

OK to freeze, desiccated (hygroscopic), protect from light


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters