Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 mg | Estimated to ship TODAYfromAreál Kühne+Nagel spol. s r.o. | CZK 7,340.00 |
About This Item
description
RTECS - TZ8335000
Quality Segment
assay
≥98% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
color
off-white
solubility
methanol: 5 mg/mL
shipped in
ambient
storage temp.
2-8°C
SMILES string
NC(C(O)CC(O)CCCCCCC(CC(OC(=O)CC(CC(=O)O)C(=O)O)C(OC(=O)CC(CC(=O)O)C(=O)O)C(CCCC)C)C)C
InChI
1S/C34H59NO14/c1-5-6-12-21(3)32(49-31(43)18-24(34(46)47)16-29(40)41)27(48-30(42)17-23(33(44)45)15-28(38)39)14-20(2)11-9-7-8-10-13-25(36)19-26(37)22(4)35/h20-27,32,36-37H,5-19,35H2,1-4H3,(H,38,39)(H,40,41)(H,44,45)(H,46,47)
InChI key
UXDPXZQHTDAXOZ-UHFFFAOYSA-N
General description
Biochem/physiol Actions
Primary rat hepatocytes
Preparation Note
Other Notes
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| form solid | form solid | form solid | form solid |
| assay ≥98% (HPLC) | assay ≥95% (HPLC) | assay ≥95% (HPLC) | assay ≥98% (TLC) |
| manufacturer/tradename Calbiochem® | manufacturer/tradename Calbiochem® | manufacturer/tradename Calbiochem® | manufacturer/tradename Calbiochem® |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| storage temp. 2-8°C | storage temp. −20°C | storage temp. 2-8°C | storage temp. 2-8°C |
| solubility methanol: 5 mg/mL | solubility DMSO: 50 mg/mL, ethanol: soluble | solubility water: 1 mg/mL | solubility DMSO: 10 mg/mL, ethanol: 5 mg/mL, acetone: soluble |
signalword
Warning
hcodes
Hazard Classifications
Carc. 2
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




