Skip to Content
Merck

Skip To

324708

Asp-N Protease

Cleaves N-terminal to aspartic acid residues, suitable for Mass Spectrometry, from Pseudomonas fragi mutant strain

Synonym(s):

Endoproteinase Asp-N, Excision Grade, Pseudomonas fragi

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
2 μg

Available to ship TODAYfromAreál Kühne+Nagel spol. s r.o.

CZK 5,770.00

About This Item

CAS Number:
EC Number:
MDL number:
NACRES:
NA.54
UNSPSC Code:
12352202
eCl@ss:
32160410
Specific activity:
≥20,000 units/mg protein
Assay:
≥90% (SDS-PAGE)

CZK 5,770.00


Available to ship TODAYDetails


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Endoproteinase Asp-N, Excision Grade, Pseudomonas fragi,

grade

Proteomics Grade

Quality Segment

assay

≥90% (SDS-PAGE)

form

lyophilized solid

specific activity

≥20,000 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: soluble

shipped in

ambient

storage temp.

2-8°C

General description

Native endoproteinase Asp-N from Pseudomonas fragi. Designed for protein sequencing or sequence verification, analysis of protein structural domains, and cleavage of fusion proteins. Metalloprotease that specifically hydrolyzes proteins at the N-terminal side of aspartic acid and cysteic acid residues. Inhibited by aprotinin, DFP, leupeptin, and TLCK. Suggested working concentration: 1:20 to 1:100 (protease:protein by weight) for sequence analysis.

Physical form

Lyophilized from 10 mM Tris-HCl, pH 7.5.

Preparation Note

Following reconstitution, aliquot and freeze (-20°C) for long term storage or refrigerate (4°C) for short term storage. Stock solutions are stable for up to 2 days at 4°C or for up to 1 month at -20°C.

Other Notes

Drapeau, G.R. 1980. J. Biol. Chem.255, 839.
One unit is defined as the amount of enzyme that will release a sufficient amount of azo dye from AZOCOLL Substrate (Cat. No. 194932) to produce an increase in absorbance at 520 nm of 0.001 per min at 37°C, pH 7.5.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Harmful (C)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P3303324715324713
grade

Proteomics Grade

grade

Proteomics Grade

grade

Proteomics Grade

grade

Proteomics Grade

specific activity

≥20,000 units/mg protein

specific activity

-

specific activity

≥200 units/mg protein

specific activity

≥10 units/mg protein

assay

≥90% (SDS-PAGE)

assay

-

assay

≥90% (SDS-PAGE)

assay

-

form

lyophilized solid

form

lyophilized powder

form

lyophilized

form

lyophilized

shipped in

ambient

shipped in

-

shipped in

ambient

shipped in

ambient

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C


Still not finding the right product?


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
324708-2UG04055977215939

Questions

Reviews

No rating value

Active Filters