Skip to Content
Merck
All Photos(1)

Documents

324519

Sigma-Aldrich

eIF4E/eIF4G Interaction Inhibitor, 4EGI-1

InSolution, ≥95%, reversibly binds eukaryotic translation initiation factor 4E

Synonym(s):

InSolution eIF4E/eIF4G Interaction Inhibitor, 4EGI-1, 2-((4-(3,4-Dichlorophenyl)-thiazol-2-ylhydrazono)-3-(2-nitrophenyl))propionic acid, eIF4F Inhibitor I, eIF4F Inhibitor I, 2-((4-(3,4-Dichlorophenyl)-thiazol-2-ylhydrazono)-3-(2-nitrophenyl))propionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H12Cl2N4O4S
CAS Number:
Molecular Weight:
451.28
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (sum of two isomers, HPLC)

form

liquid

potency

25 μM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

shipped in

dry ice

storage temp.

−70°C

InChI

1S/C18H12Cl2N4O4S/c19-12-6-5-10(7-13(12)20)15-9-29-18(21-15)23-22-14(17(25)26)8-11-3-1-2-4-16(11)24(27)28/h1-7,9H,8H2,(H,21,23)(H,25,26)/b22-14+

InChI key

KFRKRECSIYXARE-HYARGMPZSA-N

General description

A cell-permeable hydrazone compound that reversibly binds eukaryotic translation initiation factor 4E (eIF4E; KD = 25 µM) and disrupts eIF4E/eIF4G, but not eIF4E/4E-BP1, complex formation. Reported to downregulate the protein, but not mRNA, levels of c-Myc and Bcl-xL in Jurkat cells (8 hrs), leading to apoptotic cell death after prolonged exposure (24 hrs, IC50 ~25 µM). Shown to be brain permeant and reduce neuroligin protein levels, and restore L-LTP threshold and rescue autistic-like behaviours in Eif4ebp2 knockout mice.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
eIF4E
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 25 mM (5 mg/443 µL) solution of eIF4E/eIF4G Interaction Inhibitor, 4EGI-1 (Cat. No. 324517)

Reconstitution

Following initial thaw, aliquot and freeze (-70°C). Aliquots are stable for up to 6 months at -70°C.

Other Notes

Gkogkas, C.G., et al. 2012. Nature493, 371.
Moerke, N. J., et al. 2007. Cell128, 257.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reversal of memory and autism-related phenotypes in Tsc2+/- mice via inhibition of Nlgn1.
Chalkiadaki, et al.
Frontiers in Cell and Developmental Biology, 11, 1205112-1205112 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service