Skip to Content
Merck

Skip To

264157

Caspase Modulator I, 1541

Synonym(s):

Caspase Modulator I, 1541, Caspase-6 Inhibitor XI, Procaspase-3 Activator II, Procaspase-6 Activator I, 8-Methoxy-2-oxo-2H-chromene-3-carboxylic acid-(3-imidazo[1,2-a]pyridin-2-yl-phenyl)-amide, Caspase-3 Inhibitor XV

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mg

Available to ship TODAYfromAreál Kühne+Nagel spol. s r.o.

CZK 4,270.00

About This Item

Empirical Formula (Hill Notation):
C24H17N3O4
Molecular Weight:
411.41
NACRES:
NA.54
UNSPSC Code:
12352200
Assay:
≥95% (HPLC)
Form:
solid
Storage condition:
OK to freeze, protect from light

CZK 4,270.00


Available to ship TODAYDetails


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥95% (HPLC)

Quality Segment

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

yellow

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable phenylimidazopyridinyl-methoxy coumarin compound that inhibits caspase-3 and caspase-6 activities (IC50 = 35 and 38 µM, respectively) by targeting both the large and small subunit active sites. Kinetic studies with 1541 and its 8-Hydroxy analog (Cat. No. 529663 ) indicate that at suboptimal inhibitor : proenzyme molar ratio (100:1 or less), the compound does not effectively occupies both active sites of and stabilizes the proenzyme in an “on-state” conformation that facilitates the subsequent autoproteolysis or autoactivation process via the unoccupied active site of procaspase-3 and -6 (EC50 = 2.4 and 2.8 µM, respectively). Shown to effectively induce apoptosis (up to 50 µM) in various cancer cell lines (IC50 ranges from 3.8 to 8.7 µM) in a caspase-3-dependent, but p53-, caspase-8-, and Bak-independent manner. Exhibits little inhibitory effect against active caspase-7 (30 nM) even at concentrations as high as 50 µM, nor does it activate procaspase-1 or -7 even after prolonged incubations upto 4 and 24 h, respectively.

Packaging

Packaged under inert gas

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
264158400010218753
form

solid

form

solid

form

powder

form

solid

assay

≥95% (HPLC)

assay

≥95% (HPLC)

assay

≥97% (HPLC)

assay

-

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

storage condition

OK to freeze, protect from light

storage condition

OK to freeze, protect from light

storage condition

OK to freeze, protect from light

storage condition

OK to freeze, desiccated


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters