Merck
Skip To
2402-M
Stat-Matic II Plate Washer, with bottle
The StatMatic II automated plate washer is designed for use with 96-well microplates. Internal volume calibration rings allow you to adjust volumes from 100, 200, 250 & 300uL per well from a single-dispense stroke.
Synonym(s):
plate washer
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Pack SizeSKUAvailabilityPrice
About This Item
eCl@ss:
36102014
UNSPSC Code:
41104923
Manufacturer/tradename:
Chemicon®
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
manufacturer/tradename
Chemicon®
General description
Ideal for hand-held applications or for use in an automated line. The StatMatic II with a hand-held eight-port disposable manifold provides greater ease, speed, convenience, and affordability than a multi-channel pipette. The StatMatic II can be connected to a range of bottle sizes and can completely fill a microplate with one-handed operation -- in a matter of seconds.
The StatMatic II is designed for use with 96-well microplates. Internal volume calibration rings allow you to adjust volumes from 100, 200, 250, and 300uL per well from a single-dispense stroke.
The PTFE plunger guarantees smooth and accurate dispensing every time. Compatible with most reagents and chemicals, the design ensures that even protein-based reagents won′t cause sticking.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
General Lab Use