Skip to Content
Merck
All Photos(2)

Documents

238100

Sigma-Aldrich

CRM1 Inhibitor III

The CRM1 Inhibitor III controls the biological activity of CRM1. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

CRM1 Inhibitor III, (Z)-But-2-enedioic acid (4ʹ-bromophenyl)amide (3ʹʹ-chlorophenyl)amide, Chromosome Region Maintenance 1 ProteinInhibitor III, Exportin 1 Inhibitor III, XPO1 Inhibitor III

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12BrClN2O2
CAS Number:
Molecular Weight:
379.64
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.28

Quality Level

Assay

≥95% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 100 mg/mL

shipped in

wet ice

storage temp.

2-8°C

SMILES string

O=C(NC1=CC=CC(Cl)=C1)/C=C\C(NC2=CC=C(Br)C=C2)=O

General description

A cell-permeable butenediamide compound that acts as a potent, active site cysteine reactive irreversible inhibitor of CRM1/exportin 1. Shown to selectively repress agonist-mediated nuclear export of HDAC4/5, Rev and NFAT proteins in neonatal rat ventricular myocytes (NRVMs; EC50 = 2.2 and 3.5 nM for GFP-HDAC5 and GFP-HIV Rev) and suppress hypertrophic growth of NRVMs (EC50 = 52 nM for ANF expression inhibition) with negligible effect either on the phosphorylation of HDAC5 or on the kinase activities of PKD and CaMKII at 1 µM.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Monovich, L., et al. 2009. Biochim Biophys Acta.1789, 422.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service