Skip to Content
Merck

Skip To

233552-M

Cobra Venom Factor, Naja naja kaouthia

Cobra Venom Factor, Naja naja kaouthia, is a native cobra venom factor. Glycoprotein composed of three non-identical disulfide-bonded subunits of 68, 48, and 30 kDa.

Synonym(s):

Cobra Venom Anticomplementary Protein

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mg

Estimated to ship onJune 03, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 11,400.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.42
Form:
liquid
Assay:
≥95% (SDS-PAGE)

CZK 11,400.00


Estimated to ship onJune 03, 2026Details


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (SDS-PAGE)

form

liquid

specific activity

≥250 units/mL

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

foreign activity

Phospholipase, none detected

storage temp.

−70°C

General description

Native cobra venom factor from Naja naja kaouthia. Glycoprotein composed of three non-identical disulfide-bonded subunits with M.W. of 68 kDa (α), 48 kDa (β), and 30 kDa (γ). Cobra venom factor (CVF) is a structural and functional analog of cobra, as well as mammalian C3. Thus, in the presence of Factor B, Factor D, and Mg2+, CVF can form a stable CVF,Bb complex that is a C3/C5 convertase enzyme; however, the CVF,Bb complex is not susceptible to regulation by Factors H and I. Four to six µg of purified CVF are equal to 1.0 unit of functional activity as measured by the method of Cochrane, et al.

Other Notes

Bogers, W.M., et al. 1993. Eur. J. Immunol.23, 433.
Fritzinger, D.C., et al. 1992. J. Immunol. 149, 3554.
Muller, B. and Muller-Ruchholtz, W. 1987. Leuk. Res.11, 461.
Vogel, C.W. and Muller-Eberhard, H.J. 1984. J. Immunol. Methods73, 203.
Cochrane, C.G., et al. 1970. J. Immunol. 105, 55.
Ballows, M. and Cochrane, C.G. 1969. J. Immunol.103, 944.

Legal Information

Not available for sale outside the United States.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Harmful (C)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
341280341283341274-M
assay

≥95% (SDS-PAGE)

assay

≥95% (SDS-PAGE)

assay

≥90% (SDS-PAGE)

assay

≥95% (SDS-PAGE)

form

liquid

form

liquid

form

liquid

form

liquid

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

specific activity

≥250 units/mL

specific activity

-

specific activity

-

specific activity

-

storage temp.

−70°C

storage temp.

−70°C

storage temp.

−70°C

storage temp.

−70°C

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters