Skip to Content
Merck

Skip To

219010-M

Anti-Catalase Rabbit pAb

liquid, Calbiochem®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mL

Estimated to ship onJune 08, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 4,890.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Clone:
polyclonal
Species reactivity:
mouse, human
Application:
Citations:
-

CZK 4,890.00


Estimated to ship onJune 08, 2026Details


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

mouse, human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

isotype

IgG

storage temp.

−20°C

target post-translational modification

unmodified

General description

Anti-Catalase, rabbit polyclonal, recognizes the ~60-65 kDa human and mouse catalase. It is validated for ELISA, Western blotting, and immunoelectrophoresis.
Rabbit polyclonal antibody purified by ammonium sulfate precipitation and DEAE-cellulose ion exchange chromatography. Recognizes the ~60-65 kDa (monomer MW) catalase protein under reducing conditions.
Recognizes the ~60-65 kDa catalase protein.

Immunogen

purified, human erythrocytes catalase

Application

Indirect ELISA (1:128)

Immunoblotting (1:8000; see application references)

Immunoelectrophoresis (see comments)

Paraffin Sections (see application references)

Analysis Note

Positive Control
Red blood cells

Other Notes

The 256 kDa catalase protein is a tetramer of 4 identical ~60-65 kDa protein. Exhibits a single arc by immunoelectrophoresis (IEP) vs. human red cell lysate. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
204889-M219361PC41
clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

species reactivity

mouse, human

species reactivity

human

species reactivity

human

species reactivity

human

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

antibody form

purified antibody

antibody form

serum

antibody form

saturated ammonium sulfate (SAS) precipitated

antibody form

purified antibody

form

liquid

form

liquid

form

liquid

form

liquid

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters