Skip to Content
Merck

PC41

Anti-Cathepsin B (Ab-3) Rabbit pAb

liquid, Calbiochem®

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.43
UNSPSC Code:
12352203
Clone:
polyclonal
Species reactivity:
human
Application:
Citations:
6

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

dilution

(Immunoblotting (1 µg/mL)
Paraffin Sections (2.5 µg/mL, trypsin pre-treatment required))

isotype

IgG

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
219361IM27LNA61
biological source

rabbit

biological source

rabbit

biological source

mouse

biological source

mouse

antibody form

purified antibody

antibody form

saturated ammonium sulfate (SAS) precipitated

antibody form

purified antibody

antibody form

purified antibody

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

form

liquid

form

liquid

form

lyophilized

form

liquid

clone

polyclonal

clone

polyclonal

clone

CA10, monoclonal

clone

Mobu-1, monoclonal

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

General description

Anti-Cathepsin B (Ab-3), rabbit polyclonal, recognizes the ~43 kDa latent and ~31 kDa active forms of cathepsin B in breast carcinoma. It is validated for Western blotting and for paraffin sections.
Purified rabbit polyclonal antibody. Recognizes the ~43 kDa latent and the ~31 kDa active forms of cathepsin B.
Recognizes the ~43 kDa latent and the ~31 kDa active forms of cathepsin B in breast carcinoma tissues.

Immunogen

Human
purified human liver cathepsin B

Application

Immunoblotting (1 µg/ml)

Paraffin Sections (2.5 µg/ml, trypsin pre-treatment required)

Packaging

Please refer to vial label for lot-specific concentration.

Physical form

In 0.05 M sodium phosphate buffer, 0.2% gelatin.

Analysis Note

Positive Control
SK-HEP-1 concentrated conditioned medium or breast carcinoma tissue

Other Notes

Antibody should be titrated for optimal results in individual systems.
Barrett, A.J. and Kirschke, H. 1981. Methods Enzymol.80, 535.
Chan, S.J., et al. Proc. Natl. Acad. Sci. USA83, 7721.
Barrett, A.J. 1987. Trends Biochem. Sci.12, 193.
Sloane, B.F. 1990. in Seminars in Cancer Biology, ed. M. M. Gottesman. Vol. 1(2), 137.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mehrnoosh Saghizadeh et al.
Investigative ophthalmology & visual science, 46(10), 3604-3615 (2005-09-28)
To identify proteinases and growth factors abnormally expressed in human corneas of donors with diabetic retinopathy (DR), additional to previously described matrix metalloproteinase (MMP)-10 and -3 and insulin-like growth factor (IGF)-I. RNA was isolated from 35 normal, diabetic, and DR
Paola Pellegrini et al.
Autophagy, 10(4), 562-571 (2014-02-05)
Acidic pH is an important feature of tumor microenvironment and a major determinant of tumor progression. We reported that cancer cells upregulate autophagy as a survival mechanism to acidic stress. Inhibition of autophagy by administration of chloroquine (CQ) in combination
Wenqian Fang et al.
Translational research : the journal of laboratory and clinical medicine, 222, 28-40 (2020-05-22)
Nonalcoholic steatohepatitis (NASH) is a severe form of nonalcoholic fatty liver disease characterized by fat accumulation and inflammation in liver. Yet, the mechanistic insight and diagnostic and therapeutic options of NASH remain incompletely understood. This study tested the roles of
Xian Zhang et al.
iScience, 19, 607-622 (2019-08-26)
Transforming growth factor beta (TGF-β) signaling contributes to tissue fibrosis. Here we demonstrate that TGF-β enhances CatS and CatK expression but reduces CatB and CatL expression in mouse kidney tubular epithelial cells (TECs). CatS- and CatK deficiency reduces TEC nuclear
Yixing Wu et al.
Acta neuropathologica communications, 11(1), 132-132 (2023-08-15)
Cathepsin B is a cysteine protease that is implicated in multiple aspects of Alzheimer's disease pathogenesis. The endogenous inhibitor of this enzyme, cystatin B (CSTB) is encoded on chromosome 21. Thus, individuals who have Down syndrome, a genetic condition caused

Global Trade Item Number

SKUGTIN
PC41-100UG04055977224597

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service