Skip to Content
Merck

Skip To

217620

Anti-CD64 Mouse mAb (10.1)

liquid, ≥95% (SDS-PAGE), clone 10.1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μg
Check Cart for Availability
CZK 10,100.00

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Clone:
10.1, monoclonal
Species reactivity:
human
Application:
Citations:
-

CZK 10,100.00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

10.1, monoclonal

assay

≥95% (SDS-PAGE)

form

liquid

contains

0.5 mg/mL gentamycin sulfate as preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

dilution

(Flow Cytometry (10 µg/mL)
Frozen Sections
Immunoblotting
Immunoprecipitation
Neutralization Studies )

isotype

IgG1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Anti-CD64, mouse monoclonal, clone 10.1, recognizes the ~72 kDa CD64 protein (FcR1). It is validated for FC, WB, IP, IHC on frozen sections, and for neutralization studies.
Purified mouse monoclonal antibody. Recognizes the ~72 kDa CD64 (FcR1) protein.
Recognizes the ~72 kDa CD64 protein (FcR1 receptor).

Immunogen

Human
human rheumatoid synovial fluid cells/monocytes

Application

Flow Cytometry (10 µg/ml)

Frozen Sections (see comments)

Immunoblotting (see comments)

Immunoprecipitation (see comments)

Neutralization Studies (see comments)

Packaging

Please refer to vial label for lot-specific concentration.

Physical form

In 150 mM NaCl, 100 mM KCl, 50 mM sodium phosphate, pH 7.5.

Preparation Note

Following initial use, aliquot and freeze (-20°C) for long-term storage. Avoid freeze/thaw cycles.

Analysis Note

Negative Control
HPB-MLT
Positive Control
4937 cells

Other Notes

Dougherty, G.J., et al. 1987. Eur. J. Immunol.17, 1453.
The antibody is reported to block immunoglobulin binding to FcR1. FACS was performed on 5x105 cultured U-937 human tumor cells and analyzed by flow cytometry. Optimal binding concentration tested was 10 µg/ml when used with secondary reagent goat anti-mouse IgG, FITC conjugated. This antibody has also been reported to work for frozen sections, immunoblotting, and immunoprecipitation. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
CC43217605OP115
biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human

clone

10.1, monoclonal

clone

DH01, monoclonal

clone

15.2, monoclonal

clone

2A10, monoclonal

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody

isotype

IgG1

isotype

IgG1

isotype

IgG1

isotype

IgG2a

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
217620-100UG04055977202915