Skip to Content
Merck

Skip To

208923

CaMKII Inhibitor XII

Synonym(s):

CaMKII Inhibitor XII, Ca 2+/Calmodulin-Dependent Protein Kinase II Inhibitor XII

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C29H30ClN5O2S
Molecular Weight:
548.10
NACRES:
NA.28
UNSPSC Code:
12352200
Assay:
≥99% (HPLC)
Form:
powder
Storage condition:
OK to freeze, protect from light


assay

≥99% (HPLC)

Quality Segment

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

white

solubility

DMSO: 100 mg/mL

shipped in

wet ice

storage temp.

2-8°C

General description

A pyrimidinyl sulfonamide compound that acts as a potent, reversible and CaM non-competitive inhibitor of CaMKII activities (IC50 = 0.063 µM) with excellent selectivity over several kinases (IC50 = >60, 36, 11, 30 and 21 µM for CaMKIV, MLCK, p38α, Akt1 and PKC, respectively). Shown to exhibit ~25-fold greater potency towards CaMKII than the CaM-competitive inhibitor KN-93 (Cat. No. 422708 ).

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Asano, S., et al. 2010. Bioorg. Med. Chem. Lett.20, 6696.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
208923-5MG04055977219111

Questions

Reviews

No rating value

Active Filters