Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
250 μg | Estimated to ship TODAYfromAreál Kühne+Nagel spol. s r.o. | CZK 9,320.00 |
About This Item
Quality Segment
assay
≥85% (SDS-PAGE)
form
liquid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze, avoid repeated freeze/thaw cycles
impurities
IgG, IgA, IgM, C4, functionally active C5, albumin or ceruloplasmin, trace
shipped in
wet ice
storage temp.
−70°C
General description
Biochem/physiol Actions
Physical form
Preparation Note
Other Notes
Tack, B.F., et al. 1980. Methods Enzymol. 80, 64.
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| assay ≥85% (SDS-PAGE) | assay ≥95% (SDS-PAGE) | assay ≥90% (SDS-PAGE) | assay ≥95% (SDS-PAGE) |
| form liquid | form liquid | form liquid | form liquid |
| shipped in wet ice | shipped in wet ice | shipped in wet ice | shipped in wet ice |
| impurities IgG, IgA, IgM, C4, functionally active C5, albumin or ceruloplasmin, trace | impurities IgG, IgA, IgM, albumin, ceruloplasmin or functionally C3, C6, C7, C8, or C9, trace | impurities IgG, IgA, IgM, C3, C4, albumin, fibrinogen, α₂-macroglobulin, trace | impurities IgG, IgA, IgM, albumin, C3, C4, C5, C6, C7, or C8, trace |
| storage condition OK to freeze, avoid repeated freeze/thaw cycles | storage condition OK to freeze, avoid repeated freeze/thaw cycles | storage condition OK to freeze, avoid repeated freeze/thaw cycles | storage condition OK to freeze, avoid repeated freeze/thaw cycles |
| Quality Level 100 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
Storage Class
12 - Non Combustible Liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



