Skip to Content
Merck

Skip To

20-306

ATP

liquid

Synonym(s):

Adenosine triphosphate solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
300 μL

Estimated to ship TODAYfromAreál Kühne+Nagel spol. s r.o.

CZK 3,870.00

About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405
NACRES:
NA.41
Form:
liquid
Solubility:
water: soluble

CZK 3,870.00


Estimated to ship TODAYDetails


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

10mM ATP,

form

liquid

Quality Level

manufacturer/tradename

Upstate®

solubility

water: soluble

shipped in

wet ice

General description

Adenosine triphosphate (ATP) comprises an adenine base linked to sugar ribose and three phosphate groups.

Application

10mM ATP has been used in cyclin-dependent kinase 5 (CDK5) and calpain activity assay to determine CDK5 activity in brain tissues.[1]

Biochem/physiol Actions

Adenosine triphosphate (ATP) is the most crucial energy currency of cells, which is implicated in many cellular processes. It has high phosphate transfer potential, which is useful in muscle contraction, membrane transport, synthesis, and degradation of biological molecules. ATP functions as an intracellular/extracellular signaling molecule in cellular processes like cell motility, insulin secretion, neurotransmission, and organ development.[2]

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1872311051440001DATP-RO
manufacturer/tradename

Upstate®

manufacturer/tradename

-

manufacturer/tradename

Roche

manufacturer/tradename

Roche

Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

-

form

liquid

form

liquid

form

solution

form

solution

shipped in

wet ice

shipped in

-

shipped in

-

shipped in

-


Still not finding the right product?

Explore all of our products under 10mM ATP


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Phosphorus Compounds of Natural Origin: Prebiotic, Stereochemistry, Application
Kolodiazhnyi OI
Symmetry, 13(5), 889-889 (2021)
Hiromi Imamura et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(37), 15651-15656 (2009-09-02)
Adenosine 5'-triphosphate (ATP) is the major energy currency of cells and is involved in many cellular processes. However, there is no method for real-time monitoring of ATP levels inside individual living cells. To visualize ATP levels, we generated a series
Qing-Zhang Tuo et al.
Cell death & disease, 9(9), 870-870 (2018-08-31)
CDK5 activation promotes ischemic neuronal death in stroke, with the recognized activation mechanism being calpain-dependent p35 cleavage to p25. Here we reported that CDK5-Tyr15 phosphorylation by zinc induced CDK5 activation in brain ischemic injury. CDK5 activation and CDK5-Tyr15 phosphorylation were



Global Trade Item Number

SKUGTIN
20-30604053252677151

Questions

Reviews

No rating value

Active Filters