Skip to Content
Merck

Skip To

20-191

Bovine Serum Albumin

10% in TBS

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.77


Product Name

10% BSA in TBS,

form

liquid

Quality Level

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

shipped in

wet ice

storage temp.

2-8°C

Application

For use as a blocking buffer in the non-radioactive kinase plate assays.
Research Category
All

Physical form

10% BSA in TBS, pH 7.4 containing 0.05% Kathon®

Preparation Note

1 year at 4°C

Legal Information

Kathon is a registered trademark of Rohm and Haas Co.
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
20-190524750524753
technique(s)

activity assay: suitable (kinase)

technique(s)

-

technique(s)

-

technique(s)

-

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

form

liquid

form

liquid

form

-

form

-

storage temp.

2-8°C

storage temp.

-

storage temp.

15-25°C

storage temp.

15-25°C

shipped in

wet ice

shipped in

wet ice

shipped in

ambient

shipped in

ambient

manufacturer/tradename

Upstate®

manufacturer/tradename

Upstate®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®


Still not finding the right product?

Explore all of our products under 10% BSA in TBS


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
20-19104053252279010

Questions

Reviews

No rating value

Active Filters