Skip to Content
Merck

Skip To

20-155

High Salt Immune Complex Wash Buffer

For use in the Chromatin Immunoprecipitation Assay Kits.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
24 mL

Available to ship TODAYfromAreál Kühne+Nagel spol. s r.o.

CZK 4,000.00

About This Item

UNSPSC Code:
41116012
NACRES:
NA.41
eCl@ss:
32160405

CZK 4,000.00


Available to ship TODAYDetails


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

form

liquid

manufacturer/tradename

Upstate®

shipped in

wet ice

Application

For use in the Chromatin Immunoprecipitation Assay Kits.
Research Category
All

Physical form

0.1% SDS, 1% Triton X-100, 2mM EDTA, 20mM Tris-HCl, pH 8.1, 500mM NaCl.

Preparation Note

1 year at 4°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
20-15420-15620-210
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

form

liquid

form

liquid

form

liquid

form

liquid

manufacturer/tradename

Upstate®

manufacturer/tradename

Upstate®

manufacturer/tradename

Upstate®

manufacturer/tradename

Upstate®

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice


pictograms

Exclamation mark

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - ED ENV 1 - Eye Irrit. 2


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

9002-93-1

CAS No.

9002-93-1

CAS No.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



J Jason Collier et al.
American journal of physiology. Endocrinology and metabolism, 293(1), E48-E56 (2007-03-08)
Increased glucose flux generates metabolic signals that control transcriptional programs through poorly understood mechanisms. Previously, we demonstrated a necessity in hepatocytes for c-Myc in the regulation of a prototypical glucose-responsive gene, L-type pyruvate kinase (L-PK) (Collier JJ, Doan TT, Daniels
Yu-Hui Li et al.
Frontiers in oncology, 10, 567-567 (2020-05-22)
Global incidence and mortality associated with hepatocellular carcinoma (HCC) is steadily increasing. Metastasis-associated 1 (MTA1) can induce tumorigenesis and metastatic progression in HCC. However, the mechanistic details of MTA1-mediated regulation of HCC has not been completely defined. Epigenetic histone modification



Global Trade Item Number

SKUGTIN
20-15504053252622304

Questions

Reviews

No rating value

Active Filters