Skip to Content
Merck

Skip To

200965

Big CHAP - CAS 86303-22-2 - Calbiochem

Non-ionic detergent analog of CHAPS and CHAPSO with significantly reduced electrostatic interactions that do not interfere in anion exchange chromatography on DEAE-cellulose. Aggregation number: 10.

Synonym(s):

BigCHAP, N,N-Bis[3-(D-gluconamido)propyl]cholamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 g
Check Cart for Availability
CZK 7,400.00

About This Item

Empirical Formula (Hill Notation):
C42H75N3O16
CAS Number:
Molecular Weight:
878.06
UNSPSC Code:
12161900
NACRES:
NA.25
MDL number:

CZK 7,400.00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

SMILES string

C[C@H](CCC(=O)N(CCCNC(=O)[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO)CCCNC(=O)[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C

InChI

1S/C42H75N3O16/c1-21(24-7-8-25-32-26(18-30(52)42(24,25)3)41(2)11-10-23(48)16-22(41)17-27(32)49)6-9-31(53)45(14-4-12-43-39(60)37(58)35(56)33(54)28(50)19-46)15-5-13-44-40(61)38(59)36(57)34(55)29(51)20-47/h21-30,32-38,46-52,54-59H,4-20H2,1-3H3,(H,43,60)(H,44

InChI key

ZWEVPYNPHSPIFU-AUGHYPCGSA-N


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
256455-MY0000848409472
Quality Level

200

Quality Level

100

Quality Level

-

Quality Level

100


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
200965-1GM04055977221565

Questions

Reviews

No rating value

Active Filters