Skip to Content
Merck

Skip To

196878-M

(±)-Bay K 8644

Synthetic dihydropyridine derivative that acts as an active Ca2+ slow channel agonist in neuroendocrine, muscle, thyroid and other cell types.

Synonym(s):

(±)-Bay K 8644, 1,4-Dihydro-2,6-dimethyl-5-nitro-4-[2ʹ-(trifluoromethyl)phenyl]-3-pyridinecarboxylic Acid Methyl Ester

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C16H15F3N2O4
Molecular Weight:
356.30
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
solid
Storage condition:
OK to freeze, protect from light


Quality Segment

assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

yellow

solubility

DMSO: 35 mg/mL, ethanol: 35 mg/mL

storage temp.

2-8°C

General description

A synthetic dihydropyridine derivative that is an active Ca2+ slow channel agonist in neuroendocrine, muscle, thyroid, and other cell types. Prolongs single channel open time without affecting the close time. An L-type Ca2+ channel agonist. Composed of two optical isomers. The (-)-enantiomer has strong vasoconstrictive, positive ionotropic, and Ca2+ agonistic properties, whereas the (+)-enantiomer has weak vasodilating, negative ionotropic, and Ca2+ antagonistic properties. The net effect of the racemic mix is that of the negative enantiomer. Promotes β-cell proliferation/regeneration.
Synthetic dihydropyridine derivative that acts as an active Ca2+ slow channel agonist in neuroendocrine, muscle, thyroid and other cell types. Prolongs single channel open time without affecting the close time. An L-type Ca2+ channel agonist. Composed of two optical isomers. The (-)-enantiomer has strong vasoconstrictive, positive inotropic, and Ca2+ agonistic properties, whereas the (+)-enantiomer has weakly vasodilating, negative inotropic, and Ca2+ antagonistic properties. The net effect of the racemic mix is that of the negative enantiomer. Promotes β-cell proliferation/regeneration.

Biochem/physiol Actions

Primary Target
L-type Ca2+ channel

Preparation Note

Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 20 days at 4°C.

Other Notes

Wang, W., et al. 2009. Proc. Natl. Acad. Sci. USA106, 1427.
Weigl, L.G., et al. 2000. J. Physiol.525 (pt. 2), 461.
Vannier, C., et al. 1995. Am. J. Physiol. 268, L201.
Bechem, M., and Hoffmann, H. 1993. Pflugers Arch. 424, 343.
Triggle, D.J., and Rompe, D. 1989. Trends Pharmacol. Sci. 10, 507.
Takasu, N., et al. 1987. Biochem. Biophys. Res. Commun.143, 1107.
Tagliatela, M., et al. 1986. Brain Res.381, 356.
Franckowiak, G., et al. 1985. Eur. J. Pharmacol.114, 223.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Irritant (B)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
B133B112203390
form

solid

form

solid

form

powder

form

solid

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥97.5% (HPLC)

assay

≥97% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

manufacturer/tradename

Calbiochem®

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

Calbiochem®

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

solubility

DMSO: 35 mg/mL, ethanol: 35 mg/mL

solubility

H2O: slightly soluble, ethanol: soluble (Organic solutions are stable for at least 20 days at RT.)

solubility

DMSO: 20 mg/mL, methanol and ethanol: 63 mg/mL, H2O: insoluble

solubility

methanol: 1.5 mg/mL, 100% DMSO: 100 mg/mL, 90% DMSO: 75 mg/mL


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
196878-1MG04055977206609
196878-5MG04055977206616

Questions

Reviews

No rating value

Active Filters