Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
Quality Segment
description
RTECS - YV9350800
assay
≥97% (HPLC)
form
powder
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze, desiccated (hygroscopic), protect from light
color
white
solubility
water: 1 mg/mL, DMSO: 5 mg/mL
shipped in
ambient
storage temp.
2-8°C
SMILES string
[Cl-].[Cl-].N([C@@H](C(C)C)C(=O)NC(=O)[C@@H](NC(=O)NC(Cc1ccccc1)C(=O)O)CCCN=C(N)N)C(CCCN=C(N)N)C=O.[H+].[H+]
InChI
1S/C27H44N10O6.2ClH/c1-16(2)21(34-18(15-38)10-6-12-32-25(28)29)23(40)37-22(39)19(11-7-13-33-26(30)31)35-27(43)36-20(24(41)42)14-17-8-4-3-5-9-17;;/h3-5,8-9,15-16,18-21,34H,6-7,10-14H2,1-2H3,(H,41,42)(H4,28,29,32)(H4,30,31,33)(H2,35,36,43)(H,37,39,40);2*1H/t18?,19-,20?,21-;;/m0../s1
InChI key
GXQPAKIVOCLCHD-BXLPLHKWSA-N
General description
Biochem/physiol Actions
trypsin-like serine proteases, papain, and some cysteine proteases
Preparation Note
Other Notes
Ichikawa, S., et al. 1985. J. Reprod. Fertil.73, 385
Umezawa, H. 1976. Methods Enzymol.55, 678.
Suda, H., et al. 1972. J. Antibiot. (Tokyo) 25, 263.
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| description Peptidyl arginine aldehyde protease inhibitor produced by actinomycetes. | description Peptidyl arginine aldehyde protease inhibitor produced by actinomycetes. | description Alternative to DFP and PMSF. | description An enhanced water-soluble form of Bisindolylmaleimide I. |
| form powder | form solid | form solid | form solid |
| assay ≥97% (HPLC) | assay - | assay ≥97% (HPLC) | assay ≥95% (HPLC) |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| manufacturer/tradename Calbiochem® | manufacturer/tradename Calbiochem® | manufacturer/tradename Calbiochem® | manufacturer/tradename Calbiochem® |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
| storage condition desiccated (hygroscopic), OK to freeze, protect from light | storage condition OK to freeze, desiccated (hygroscopic) | storage condition OK to freeze, desiccated | storage condition OK to freeze |
Still not finding the right product?
Explore all of our products under Antipain, Dihydrochloride
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


