Select a Size
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 25 assays | Estimated to ship onMay 29, 2026fromAreál Kühne+Nagel spol. s r.o. | CZK 14,300.00 |
About This Item
General description
SUBUNIT STRUCTURE: Interacts with SIN3A, SIN3B and RCOR1.
SUBCELLULAR LOCATION: NucleusProbable.
TISSUE SPECIFICITY: Ubiquitous. Expressed at higher levels in the tissues of the lymphocytic compartment, including spleen, thymus, peripheral blood lymphocytes and ovary.
SEQUENCE SIMILARITY:Contains 9 C2H2-type zinc fingers.
SEQUENCE CAUTION: The sequence AAA98503.1 differs from that shown. Reason: Frameshift at positions 189, 212 and 1087.
The sequence AAH38985.1 differs from that shown. Reason: Miscellaneous discrepancy. Contaminating sequence. Potential poly-A sequence.
Application
Biochem/physiol Actions
Packaging
Physical form
Normal Rabbit IgG. One vial containing 125 μg purified rabbit IgG in 125 μL storage buffer containing 0.1% sodium azide. Store at -20°C.
ChIP Primers, Synapsin RE1. One vial containing 75 μL of 5 μM of each primer specific for the RE1 site of human Synapsin. Store at -20°C.
FOR: GGT GCT GAA GCT GGC AGT
REV: TGG GTT TTA GGA CCA GGA TG
Preparation Note
Analysis Note
Sonicated chromatin prepared from HeLa S3 cells (4 x 106 cell equivalents per IP) were subjected to chromatin immunoprecipitation using 2 µg of either a normal rabbit IgG or Anti-REST antibody and the Magna ChIP A Kit (Part No. 17-610) (Please see figures). Successful immunoprecipitation of REST-associated DNA fragments was verified by qPCR using Control Primers.
Other Notes
Disclaimer
Still not finding the right product?
Explore all of our products under ChIPAb+ REST - ChIP Validated Antibody and Primer Set
Storage Class
10 - Combustible liquids
wgk
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Signaling Product Guide: Antibodies, small molecule inhibitors, kits, assays and proteins for signaling research.
Epigenetics describes heritable changes in gene expression caused by non-genetic mechanisms. Epigenetic regulation allows a cell to vary its response based on its biological and environmental contexts. Epigenetic changes can effect transcriptional and post-transcriptional regulation via mechanisms such as histone modification, chromatin and nucleosome remodeling, DNA methylation, and small and non-coding RNA-mediated regulation. These mechanisms, in cooperation with transcription factors and other nucleic acid-binding proteins, regulate gene expression. Epigenetic mechanisms of gene regulation impacts diverse areas of research—from agriculture to human health. Common epigenetic assays such as chromatin immunoprecipitation (ChIP) and RNA immunoprecipitation (RIP) rely on high quality antibodies that recognize specific epigenetic modifications for accurate results. EMD Millipore offers over 100 ChIPAb+™ and RIPAb+™ validated antibody kits that are quality tested on ChIP/RIP assays and are conveniently provided with control qPCR primers and negative control antibodies to ensure first time ChIP/RIP success.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 17-641 | 04053252271496 |