Skip to Content
Merck

Skip To

17-609

ChIPAb+ Acetyl-Histone H3 (Lys9) Serum - ChIP Validated Antibody and Primer Set

serum, from rabbit

Synonym(s):

H3K9Ac, Histone H3 (acetyl K9)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
25 assays

Estimated to ship onMay 07, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 13,900.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.52
eCl@ss:
32160702
Clone:
polyclonal
Species reactivity:
human, mouse
Application:
ChIP, IP, WB
Citations:
4

CZK 13,900.00


Estimated to ship onMay 07, 2026Details


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

antibody form

serum

clone

polyclonal

species reactivity

human, mouse

species reactivity (predicted by homology)

mammals

manufacturer/tradename

ChIPAb+, Upstate®

technique(s)

ChIP: suitable, immunoprecipitation (IP): suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... H3F3B(3021)

General description

17 kDa
All ChIPAb+ antibodies are individually validated for chromatin precipitation, every lot, every time. Each ChIPAb+ antibody set includes control primers (tested every lot by qPCR) to biologically validate your IP results in a locus-specific context. The qPCR protocol and primer sequences are provided, allowing researchers to validate ChIP protocols when using our antibody in their chromatin context. Each set also includes a negative control antibody to ensure specificity of the ChIP reaction.
The ChIPAb+ Acetyl-Histone H3 (Lys9) set includes the anti-acetyl-histone H3 (Lys9) antibody, a negative control antibody (normal rabbit serum), and qPCR primers which amplify a 166 base pair region within the promoter of the human GAPDH gene. The acetyl-histone H3 (Lys9) and negative control antibodies are supplied in a scalable "per ChIP" reaction size and can be used to functionally validate the precipitation of acetyl-histone H3 (Lys9) associated chromatin.

Immunogen

The acetyl-histone H3 (Lys9) antiserum is made against a peptide corresponding to amino acids 4-14 of yeast histone H3 acetylated on lysine 9.

Application

Chromatin Immunoprecipitation:
Sonicated chromatin prepared from untreated HeLa cells (1 X 106 cell equivalents) was subjected to chromatin immunoprecipitation using 1 μL of either a normal rabbit antiserum or Anti-Acetyl-Histone H3 (Lys9) serum and the Magna ChIP A Kit (Cat. #17-610) (Figure 2). Successful immunoprecipitation of acetyl-histone H3 (Lys9) associated DNA fragments was verified by qPCR using control ChIP Primers flanking the human GAPDH promoter or primers amplifying the promoter of human β-globin, which is transcriptionally inactive in HeLa cells. Percent Input relative to standard curves for each qPCR primer set are shown, with immunoprecipitated DNA from control serum shown as (-) and acetylhistone H3 serum as (+).
Please refer to the EZ-Magna A ChIP (Cat. #17-408) or EZ-ChIP (Cat. #17-371) protocol for experimental details.

Western Blot Analysis:
Acid-extracted proteins from normal HeLa cells (Lane 1) and HeLa cells treated with 5mM sodium butyrate for 24 hours (Lane 2) were resolved by electrophoresis, transferred to PVDF membrane and probed with Anti-Acetyl-Histone H3 (Lys9) (1:5000). Proteins were visualized using a goat-anti rabbit secondary antibody conjugated to HRP and chemiluminescent detection system (Please see figures).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology
This ChIPAb+ Acetyl-Histone H3 (Lys9) Serum -ChIP Validated Antibody & Primer Set conveniently includes the antibody & the specific control PCR primers.

Biochem/physiol Actions

Acetyl-Histone H3 (Lys9)

Packaging

25 assays per kit, ~1μL per chromatin immunoprecipitation

Physical form

Anti-Acetyl-Histone H3 (Lys9) (rabbit polyclonal serum). One vial containing 25 μL of antiserum containing 0.05% sodium azide before the addition of glycerol to 30%.

Normal Rabbit Serum. One vial containing 25 uL antiserum containing 0.05% sodium azide.

Control Primers. One vial containing 75 μL of 5 μM of each primer specific for for human GAPDH.
FOR: TAC TAG CGG TTT TAC GGG CG
REV: TCG AAC AGG AGG AGC AGA GAG CGA

Preparation Note

Stable for 1 year at -20°C from date of receipt

Analysis Note

Chromatin Immunoprecipitation:
Sonicated chromatin prepared from untreated HeLa cells (1 X 106 cell equivalents) was subjected to chromatin immunoprecipitation using 1 μL of either a normal rabbit antiserum or Anti-Acetyl-Histone H3 (Lys9) serum and the Magna ChIP A (Cat. #17-610) Kit.
Successful immunoprecipitation of acetylhistone H3 (Lys9) associated DNA fragments was verified by qPCR using control ChIP Primers flanking the human GAPDH promoter (Please see figures).
Please refer to the EZ-Magna A ChIP (Cat. #17-408) or EZ-ChIP (Cat. #17-371) protocol for experimental details.
Control
Included negative control antibody normal rabbit serum and control primers specific for human GAPDH promoter.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
17-65817-68017-681
clone

polyclonal

clone

polyclonal

clone

CMA306, monoclonal

clone

CMA307, monoclonal

biological source

rabbit

biological source

rabbit

biological source

mouse

biological source

mouse

species reactivity

human, mouse

species reactivity

human, mouse

species reactivity

vertebrates, human

species reactivity

human, vertebrates

antibody form

serum

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

-

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

Gene Information

human ... H3F3B(3021)

Gene Information

human ... H3F3B(3021)

Gene Information

-

Gene Information

-


Storage Class

10 - Combustible liquids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Epigenetics describes heritable changes in gene expression caused by non-genetic mechanisms. Epigenetic regulation allows a cell to vary its response based on its biological and environmental contexts. Epigenetic changes can effect transcriptional and post-transcriptional regulation via mechanisms such as histone modification, chromatin and nucleosome remodeling, DNA methylation, and small and non-coding RNA-mediated regulation. These mechanisms, in cooperation with transcription factors and other nucleic acid-binding proteins, regulate gene expression. Epigenetic mechanisms of gene regulation impacts diverse areas of research—from agriculture to human health. Common epigenetic assays such as chromatin immunoprecipitation (ChIP) and RNA immunoprecipitation (RIP) rely on high quality antibodies that recognize specific epigenetic modifications for accurate results. EMD Millipore offers over 100 ChIPAb+™ and RIPAb+™ validated antibody kits that are quality tested on ChIP/RIP assays and are conveniently provided with control qPCR primers and negative control antibodies to ensure first time ChIP/RIP success.


Keisuke Mori et al.
PloS one, 9(1), e87319-e87319 (2014-02-06)
We previously reported that sevoflurane anesthesia reversibly suppresses the expression of the clock gene, Period2 (Per2), in the mouse suprachiasmatic nucleus (SCN). However, the molecular mechanisms underlying this suppression remain unclear. In this study, we examined the possibility that sevoflurane
Raffaella Gatta et al.
Cell cycle (Georgetown, Tex.), 9(11), 2149-2159 (2010-05-28)
Histones are modified by different post-translational modifications which are marks of peculiar chromatin functions. We previously evaluated histone methylations of G1/S and G2/M cell-cycle promoters at the single nucleosome level; here we report an analysis of acetylation marks, including some
Qingsong Zhu et al.
Amino acids, 42(2-3), 887-898 (2011-08-02)
Aberrant epigenetic repression of gene expression has been implicated in most cancers, including breast cancer. The nuclear amine oxidase, lysine-specific demethylase 1 (LSD1) has the ability to broadly repress gene expression by removing the activating mono- and di-methylation marks at



Global Trade Item Number

SKUGTIN
17-60904053252587788

Questions

Reviews

No rating value

Active Filters