Skip to Content
Merck
All Photos(1)

Key Documents

17-439

Sigma-Aldrich

RalB Activation Assay Kit (non-radioactive)

Non-radioactive Ral-B Activation Assay Kit can be used to precipitate Ral-B-GTP from cell lysates & detection by a Ral-B specific monoclonal antibody.

Synonym(s):

RAS-like protein B, v-ral simian leukemia viral oncogene homolog B, GTP binding protein

Sign Into View Organizational & Contract Pricing

Select a Size

20 ASSAYS
CZK 26,000.00

CZK 26,000.00


Estimated to ship onMarch 06, 2025Details



Select a Size

Change View
20 ASSAYS
CZK 26,000.00

About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

CZK 26,000.00


Estimated to ship onMarch 06, 2025Details


Quality Level

species reactivity

mouse, rat, human

manufacturer/tradename

Upstate®

technique(s)

affinity binding assay: suitable (G-protein)

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Components

anti-RalB, clone 25 (Cat.# 04-037)

Ral Assay Reagent (Ral BP1, agarose) (Cat.# 14-415)

Mg2+ Lysis/Wash Buffer, 5X (Cat.# 20-168)

100X GTPγS, 10mM (Cat.# 20-176)

100X GDP, 100mM (Cat.# 20-177)

Quality

Routinely evaluated by non-radioactive Kinase Assay.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service