Select a Size
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 100 mL | Please contact Customer Service for Availability | CZK 5,710.00 |
About This Item
grade
certified reference material
Quality Segment
product line
Certipur®
technique(s)
ICP: suitable
pH
0.5 (20 °C in H2O)
density
1.013 g/cm3 at 20 °C
application(s)
industrial qc
pharmaceutical
format
single component solution
storage temp.
15-25°C
General description
Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.
Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty, date of expiry, and detailed information about trace impurities.
Application
Analysis Note
Other Notes
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) ICP: suitable | technique(s) ICP: suitable | technique(s) ICP: suitable | technique(s) ICP: suitable |
| grade certified reference material | grade certified reference material | grade certified reference material, TraceCERT® | grade certified reference material, TraceCERT® |
| format single component solution | format single component solution | format single component solution | format single component solution |
| product line Certipur® | product line Certipur® | product line TraceCERT® | product line TraceCERT® |
| application(s) industrial qc | application(s) industrial qc | application(s) industrial qc | application(s) industrial qc |
| storage temp. 15-25°C | storage temp. 15-25°C | storage temp. - | storage temp. - |
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2
Storage Class
12 - Non Combustible Liquids
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 1703670100 | 04022536739090 |



