Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
UNSPSC Code:
41106227
Pricing and availability is not currently available.
material
polystyrene plate
Quality Segment
agency
EP, USP
sterility
sterile; γ-irradiated
form
plate
feature
2D bar code, ready-to-use
packaging
pkg of (20 Plates per box)
D
90 mm
pH
(7.3 (in H2O))
application(s)
air monitoring
microbiology
pharmaceutical
storage temp.
15-25°C
suitability
(Yeasts and Molds)
General description
Sabouraud Dextrose Agar + LT - ICR plus settle plate is a selective media for the determination of the total count of yeasts and molds. Settle plates are used for active or passive air monitoring as well for fingerprints of personnel. Sabouraud Dextrose Medium contains digests of animal tissues (peptones) which provide a nutritious source of amino acids and nitrogenous compounds for the growth of fungi and yeasts. Dextrose is added as the energy and carbon source and low pH favours fungal growth. The medium is supplemented with Lecithin and polysorbate 80 (Tween®) as neutralizers to inactivate residues of disinfectants. In addition, the plate model (plus or “+”) is supplied with a lockable lid for safe transport of the samples without the risk of losing the lid. For anaerobic incubation the lid needs to be turned into the vent position.
Application
Sabouraud Dextrose Agar (SDA) is a selective medium for cultivation and isolation of yeasts and molds. The media is used for environmental monitoring by determining the yeast and mold count in air by active or passive air monitoring as well as fingerprints of personnel.
Packaging
1667030020-CN: 20 Plates per box
Legal Information
TWEEN is a registered trademark of Croda International PLC
Still not finding the right product?
Explore all of our products under Sabouraud Dextrose Agar
Storage Class
13 - Non Combustible Solids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.