Skip to Content
Merck
All Photos(1)

Key Documents

14-778-M

Sigma-Aldrich

PP5, active

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
CZK 11,100.00
250 μG
CZK 89,400.00

CZK 11,100.00


Estimated to ship onApril 07, 2025Details


Request a Bulk Order

Select a Size

Change View
10 μG
CZK 11,100.00
250 μG
CZK 89,400.00

About This Item

UNSPSC Code:
12352204
eCl@ss:
32160405
NACRES:
NA.32

CZK 11,100.00


Estimated to ship onApril 07, 2025Details


Request a Bulk Order

biological source

human

Quality Level

mol wt

Mw 83.8 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (phosphatase)

NCBI accession no.

Gene Information

human ... PPP5C(5536)

General description

Product Source: N-terminal GST-tagged, recombinant, full length, human PP5, expressed in E. coli cells.
This protein is a serine/threonine phosphatase which is a member of the protein phosphatase catalytic subunit family.
Proteins in this family participate in pathways regulated by reversible phosphorylation at serine and threonine
residues; many of these pathways are involved in the regulation of cell growth and differentiation. The product of
this gene has been shown to participate in signaling pathways in response to hormones or cellular stress, and elevated levels of this protein may be associated with breast cancer development. Alternative splicing results in multiple transcript variants.

Packaging

Also available in 250μg size as catalog # 14-778M and in bulk as catalog # 14-778-K.

Quality

Routinely evaluated by the release of phosphate from the substrate 6,8-difluoro-4-methylumbelliferyl phosphate (DiFMUP)

Physical form

Purified using glutathione agarose.

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service