Skip to Content
Merck

Skip To

14-746-M

Src (1-530) Protein, active, 10 µg

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 μg

Estimated to ship onJune 03, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 14,300.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32
eCl@ss:
32160405
Biological source:
human
Mol wt:
Mw 61.7 kDa

CZK 14,300.00


Estimated to ship onJune 03, 2026Details


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

biological source

human

mol wt

Mw 61.7 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

GenBank accession no.

UniProt accession no.

Gene Information

human ... SRC(6714)

General description

N-terminal 6His-tagged, recombinant, human Src, amino acids 1-530, expressed by baculovirus in Sf21 insect cells.
Product Source: Expressed by baculovirus in Sf21 insect cells.
The mutant form of Src (Src, 1-530) has been identified in cases of advanced colon cancer. This truncating mutation generates an open active form of Src that has been found to be transforming, tumorigenic and metastasis promoting.

Application

Active, N-terminal 6His-tagged, recombinant, human Src, amino acids 1-530, expressed by baculovirus in Sf21 insect cells. For use in Kinase Assays.
Research Category
Metabolism
Research Sub Category
Cardiovascular Disease

Metabolic Disorders

Biochem/physiol Actions

Protein Target: Src
Target Sub-Family: TK

Packaging

Also available in 250μg size as catalog # 14-746M - Call for pricing and availability.

Physical form

50mM Tris-HCl pH7.5, 300mM NaCl, 0.1mM EGTA, 0.03% Brij-35, 270mM sucrose, 1mM benzamidine, 0.2mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution
Ni2+/NTA-agarose

Preparation Note

6 months at -70°C from date of shipment.

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
14-748-M14-326-M14-527-M
biological source

human

biological source

human

biological source

human

biological source

human

technique(s)

activity assay: suitable (kinase)

technique(s)

activity assay: suitable (kinase)

technique(s)

activity assay: suitable (kinase)

technique(s)

activity assay: suitable (kinase)

mol wt

Mw 61.7 kDa

mol wt

Mw 61.7 kDa

mol wt

Mw 61.7 kDa

mol wt

Mw 53.4 kDa

Gene Information

human ... SRC(6714)

Gene Information

human ... SRC(6714)

Gene Information

human ... SRC(6714)

Gene Information

-

UniProt accession no.

P05480 (mouse), Q9WUD9 (rat), P12931

UniProt accession no.

P12931

UniProt accession no.

P12931

UniProt accession no.

P08922

GenBank accession no.

K03218

GenBank accession no.

-

GenBank accession no.

-

GenBank accession no.

-


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters