Skip to Content
Merck

Skip To

1.46802

Tryptic Soy Broth with Neutralizers

Tween® 0.5 %, bottle capacity 500 mL, bottle filling volume 400 mL, closure type, blue screw cap with septum, box of 6 bottles

Synonym(s):

Casein Soybean Digest Broth, TSB, TSB with 0.5% Tween® 80, TSB with neutralizer, Tryptone Soy Broth, Tryptone Soy Broth + 0.5% Tween® 80

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41106225
NACRES:
NA.72
Bottle capacity:
500 mL
Bottle filling volume:
400 mL
Feature:
closure type blue screw cap with septum, ready-to-use
Agency:
EP, JP (4.06), USP
Ph:
7.1-7.5
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

EP, JP (4.06), USP

sterility

sterile; autoclaved (tested for 14 days at 20-25°C)

sterilization compatibility

autoclavable compatible

form

liquid

shelf life

360 days

feature

closure type blue screw cap with septum, ready-to-use

composition

Dipotassium Hydrogen Phosphate, 2.5 g , Glucose Monohydrate, 2.5 g , Pancreatic Digest of Casein, 17 g , Papaic Digest of Soya Bean Meal, 3 g , Polysorbate 80, 5 mL , Sodium Chloride, 5 g , Tween®, 0.5%

packaging

box of 6 bottles

bottle capacity

500 mL

bottle filling volume

400 mL

pH

7.1-7.5

application(s)

food and beverages
pharmaceutical
sterility testing

components

tween

storage temp.

2-25°C

suitability

nonselective for

General description

Tryptic Soy Broth is a nutrient-rich universal medium free of inhibitors and targets to enumerate a broad spectrum of bacteria, yeast, and molds. The medium is supplemented with peptones from casein and soymeal to provide an enhanced carbon source that promotes growth for the microflora. The broth medium is widely used during sterility testing due to its wide range of specificity. Tween® 80 in the media formulation inhibits active agents of disinfectants and promotes microbial growth present in the test sample.

Application

Tryptic Soy Broth with 0.5% Tween® 80 is used for the cultivation and enumeration of a broad range of bacteria, yeasts, and fungi in the presence of disinfectant residue. The medium is suitable for the testing of sterile and non-sterile substances, preparations, and products.

Features and Benefits

Our ready-to-use media provide the highest level of quality and testing confidence. They have been formulated and tested to meet the pharmacopeia requirements. Sterility testing media and rinse solutions are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility, and growth promotion tests. Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, thus improving accuracy and reducing the risk of incorrect interpretation and false results.

Other Notes

Production will take place at Millipore SAS in Molsheim, France. Deliveries will be executed in accordance with the terms and conditions of MLS RTU GmbH. A minimum order quantity is required to process the order. Once the order is placed and confirmed, Merck will ensure product delivery within a maximum timeframe of 10 to 12 weeks. The terms and conditions of Merck KGaA apply to both Millipore SAS and MLS RTU GmbH and can be accessed through the following link: Standard Terms & conditions
This medium can be modified and/or supplemented based on the required performance criteria.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Disclaimer

This item is created at the customer′s request as a customized product and will not be developed or qualified in accordance with the Merck Product Development Process.


Still not finding the right product?


wgk

WGK 2

Storage Class

10 - Combustible liquids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters