Skip to Content
Merck

Skip To

1.46571

Buffered Sodium Chloride Peptone Solution with Neutralizers

Tween®, Histidine, Lecithin 3 g, bottle capacity 250 mL, bottle filling volume 200 mL, closure type, Blue screw cap with 3 loci, pack of 6 bottles

Synonym(s):

Buffered NaCl Peptone Solution + LTH, Buffered Saline Peptone Water, NaCl Peptone Buffer

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352201
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

EP 2.6.1, JP 4.06, USP 71

sterility

sterile; autoclaved

form

liquid

feature

ready-to-use, closure type Blue screw cap with 3 loci

composition

Histidine , Lecithin, 3 g , Tween®

packaging

pack of 6 bottles

technique(s)

direct inoculation: suitable, microbiological culture: suitable

bottle capacity

250 mL

bottle filling volume

200 mL

pH

7.0±0.2

application(s)

cosmetics
food and beverages
pharmaceutical
sterility testing

storage temp.

2-25°C

suitability

nonselective for

General description

Buffered NaCl Peptone Solution with LTH (lecithin, Tween®80 and histidine) has a low content of peptone, along with phosphate buffer and sodium chloride, to maintain the viability of microorganisms present in test sample without supporting their growth.

Lecithin and Polysorbate 80, is effective against quaternary ammonium compounds, amphoteric surfactants, benzamidines, chlorhexidines and dequadin. Polysorbate 80 inactivates benzyl alcohol, carbanilides, dichlorbenzyl alcohols, benzoic acid, p-hydroxybenzoic acid and its esters, phenols, phenylethyl alcohols and PHB esters. Histidine is effective against formaldehyde and formaldehyde releasing agents.

Application

Buffered NaCl Peptone Solution with LTH (lecithin, Tween® 80 and histidine) is used for the microbial examination of non-sterile samples. It is suitable for dilution, sample preparation and sample washing.

Features and Benefits

  • Our ready-to-use media provide the highest level of quality and testing confidence. They have been formulated and tested to meet the pharmacopoeia requirements.
  • Sterility testing media and rinse solutions are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility, and growth promotion tests.
  • Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, therefore improving accuracy, and significantly reducing the risk of incorrect interpretation and false results.

Other Notes

Production will take place at Millipore SAS in Molsheim, France. Deliveries will be executed in accordance with the terms and conditions of MLS RTU GmbH. A minimum order quantity is required to process the order. Once the order is placed and confirmed, Merck will ensure product delivery within a maximum timeframe of 10 to 12 weeks. The terms and conditions of Merck KGaA apply to both Millipore SAS and MLS RTU GmbH and can be accessed through the following link: Standard Terms & Conditions
This medium can be modified and/or supplemented based on the required performance criteria.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Disclaimer

This item is created at the customer′s request as a customized product and will not be developed or qualified in accordance with the Merck Product Development Process.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.465071.465701.46175
technique(s)

direct inoculation: suitable, microbiological culture: suitable

technique(s)

microbiological culture: suitable, sterile filtration: suitable

technique(s)

direct inoculation: suitable, microbiological culture: suitable

technique(s)

direct inoculation: suitable, sterile filtration: suitable

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300

application(s)

cosmetics
food and beverages
pharmaceutical
sterility testing

application(s)

cosmetics
food and beverages
pharmaceutical
sterility testing

application(s)

cosmetics
food and beverages
pharmaceutical
sterility testing

application(s)

bioburden testing
cosmetics
food and beverages
pharmaceutical

form

liquid

form

liquid

form

liquid

form

liquid

storage temp.

2-25°C

storage temp.

2-25°C

storage temp.

2-25°C

storage temp.

2-25°C

pH

7.0±0.2

pH

6.8-7.2

pH

-

pH

7.0


Still not finding the right product?


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters