Select a Size
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 6 ea | Check Cart for Availability | CZK 1,250.00 |
About This Item
agency
EP 2.6.13
Quality Level
description
For the microbiological examination of non-sterile pharmaceuticals
sterility
sterile; autoclaved
form
liquid
shelf life
12 mo.
feature
closure type White screw cap for wide neck, ready-to-use
composition
Lecithin, 3 g , Sodium Thiosulfate, 5 g
packaging
pack of 6 bottles, pkg of (90 ml in 275 ml wide mouth bottle (6 bottles per box))
technique(s)
direct inoculation: suitable, sterile filtration: suitable
bottle capacity
275 mL
bottle filling volume
90 mL
pH
7.0
application(s)
bioburden testing
cosmetics
food and beverages
pharmaceutical
storage temp.
2-25°C
suitability
nonselective for
General description
Application
Features and Benefits
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) direct inoculation: suitable, sterile filtration: suitable | technique(s) direct inoculation: suitable, sterile filtration: suitable | technique(s) direct inoculation: suitable, sterile filtration: suitable | technique(s) direct inoculation: suitable, sterile filtration: suitable |
| Quality Level 300 | Quality Level 300 | Quality Level 300 | Quality Level 300 |
| application(s) bioburden testing | application(s) bioburden testing | application(s) bioburden testing | application(s) bioburden testing |
| form liquid | form liquid | form liquid | form liquid |
| storage temp. 2-25°C | storage temp. 2-25°C | storage temp. 2-30°C | storage temp. 2-25°C |
| shelf life 12 mo. | shelf life 12 mo. | shelf life 12 mo. | shelf life 12 mo. |
Still not finding the right product?
Explore all of our products under Buffered Sodium Chloride Peptone Solution with Neutralizers
Storage Class
10 - Combustible liquids
wgk
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 1464710006 | 04027269265328 |


