Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 pkg | Estimated to ship onJune 04, 2026fromAreál Kühne+Nagel spol. s r.o. | CZK 27,200.00 |
About This Item
Quality Segment
agency
EP, JP, USP
sterility
sterile; sterile-filtered
form
liquid (ready-to-use)
feature
ready-to-use
packaging
pkg of 10 L bag with MPC connector
technique(s)
cell counting: suitable (total microorganisms), media fill: suitable
pH
7.3 (20.0 °C in H2O)
application(s)
aseptic process simulation
pharmaceutical
compatibility
stand alone
storage temp.
2-25°C
suitability
suitable for (Mediafill testing), nonselective for
General description
Application
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| suitability suitable for (Mediafill testing), nonselective for | suitability - | suitability - | suitability - |
| technique(s) cell counting: suitable (total microorganisms), media fill: suitable | technique(s) media fill: suitable | technique(s) microbiological culture: suitable | technique(s) media fill: suitable, microbiological culture: suitable |
| form liquid (ready-to-use) | form liquid | form granular | form granular |
| agency EP, USP, JP | agency EP, JP, USP | agency EP, USP | agency EP, JP, USP |
| application(s) aseptic process simulation | application(s) aseptic process simulation | application(s) microbiology, pharmaceutical | application(s) aseptic process simulation, pharmaceutical |
| Quality Level 300 | Quality Level 300 | Quality Level 200 | Quality Level 400 |
Still not finding the right product?
Explore all of our products under Vegetable Peptone Broth
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Storage Class
12 - Non Combustible Liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



