Skip to Content
Merck
All Photos(1)

Documents

14-618-M

Sigma-Aldrich

CD45 Protein, 10 µg

His6-tagged, recombinant, human CD45, amino acids 598-end, for use in Enzyme Assays.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

biological source

human

Quality Level

100
300

recombinant

expressed in E. coli

mol wt

Mw 84 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable

UniProt accession no.

Gene Information

human ... PTPRC(5788)

General description

His6-tagged, recombinant, human CD45, amino acids 598-end
Product Source: expressed in E. coli

Application

Research Category
Inflammation & Immunology

Biochem/physiol Actions

Protein Target: CD45
Target Sub-Family: Tyr phosphatase

Quality

routinely evaluated by dephosphorylation of 100μM DiFMUP

Physical form

Ni2+/NTA-agarose

Storage and Stability

6 months at -70°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service