Skip to Content
Merck

Skip To

264155

Caspase-3 Inhibitor II

≥90% (TLC), irreversible CPP-32/apopain inhibitor, solid

Synonym(s):

Caspase-3 Inhibitor II, Z-D(OMe)E(Ome)VD(OMe)-FMK

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
250 μg

Estimated to ship onMay 29, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 3,580.00
1 mg

Estimated to ship onMay 29, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 10,300.00

Estimated to ship onJune 02, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 468,800.00

About This Item

Empirical Formula (Hill Notation):
C30H41FN4O12
CAS Number:
Molecular Weight:
668.66
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥90% (TLC)
Form:
solid
Storage condition:
OK to freeze, desiccated

CZK 3,580.00


Estimated to ship onMay 29, 2026Details


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Caspase-3 Inhibitor II, The Caspase-3 Inhibitor II, also referenced under CAS 210344-95-9, controls the biological activity of Caspase-3. This small molecule/inhibitor is primarily used for Cancer applications.

Quality Segment

assay

≥90% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated

color

white to slightly brown

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

FCC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)OCc1ccccc1)CC(=O)OC)CCC(=O)OC)C(C)C)CC(=O)OC

InChI

1S/C30H41FN4O12/c1-17(2)26(29(42)33-20(22(36)15-31)13-24(38)45-4)35-27(40)19(11-12-23(37)44-3)32-28(41)21(14-25(39)46-5)34-30(43)47-16-18-9-7-6-8-10-18/h6-10,17,19-21,26H,11-16H2,1-5H3,(H,32,41)(H,33,42)(H,34,43)(H,35,40)/t19-,20-,21-,26-/m0/s1

InChI key

GBJVAVGBSGRRKN-JYEBCORGSA-N

General description

A potent, cell-permeable and irreversible inhibitor of CPP-32/apopain, a member of the ICE/CED-3 family of cysteine proteases.
A potent, cell-permeable, and irreversible inhibitor of caspase-3 as well as caspase-6, caspase-7, caspase-8, and caspase-10. When using with purified native or recombinant enzyme, pretreatment with an esterase is required.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
caspase-3, caspase-6, caspase-7, caspase-8, caspase-10
Product does not compete with ATP.
Reversible: no

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Thornberry, N.A., and Lazebnik, Y. 1998. Science 281, 1312.
Masuda, Y., et al. 1997. Biochem. Biophys. Res. Commun. 234, 641.
Nicholson, D.W. 1996. Nature Biotech. 14, 297.
Schlegel, J., et al. 1996. J. Biol. Chem. 271, 1841.
Nicholson, D.W., et al. 1995. Nature 376, 37.
Z-Asp(OCH₃)-Glu(OCH₃)-Val-Asp(OCH₃)-FMK

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
400012218753218759
assay

≥90% (TLC)

assay

≥95% (HPLC)

assay

-

assay

-

form

solid

form

solid

form

solid

form

powder

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

Quality Level

200

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage condition

OK to freeze, desiccated

storage condition

OK to freeze, desiccated

storage condition

OK to freeze, desiccated

storage condition

OK to freeze, desiccated


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
264155-5UNMG04055977198461
264155-1MG04055977198447
264155-250UG04055977198454

Questions

Reviews

No rating value

Active Filters