Skip to Content
Merck

Skip To

1.46161

DG 18 Agar

ReadyPlate, plate diam. 90 mm, ready-to-use, sterile, box of 20 plates, for yeasts, for molds

Synonym(s):

Dichloran 18% Glycerol Agar, ReadyPlate DG18 Agar ISO21527

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
20 pkg

Estimated to ship TODAYfromAreál Kühne+Nagel spol. s r.o.

CZK 4,000.00

About This Item

NACRES:
NA.74
UNSPSC Code:
41106227
Plate diam.:
90 mm
Feature:
ready-to-use
Sterility:
sterile

CZK 4,000.00


Estimated to ship TODAYDetails


Request more information
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

ISO 21527

sterility

sterile

product line

ReadyPlate

form

plate (ready-to-use)

feature

ready-to-use

packaging

box of 20 plates

manufacturer/tradename

ReadyPlate

storage condition

protect from light (Store dry and tightly closed)

plate diam.

90 mm

color

clear light yellow

pH

5.6 ( in H2O)

application(s)

food and beverages
industrial qc
microbiology

storage temp.

15-25°C

suitability

molds, yeasts

General description

ReadyPlate DG18 Agar prevents the growth of bacteria by reducing the water activity from approx. 0.99 to 0.95 with 18 % (w/w) glycerol and the addition of chloramphenicol. The inclusion of dichloran inhibits the rapid spreading of mucoraceous fungi and restricts colony sizes of other genera, easing the colony count. The enzymatic digest of casein is a nitrogen source containing a high level of free amino acids. Glucose provides an energy source for growth, while agar is the solidifying agent.

Application

ReadyPlateDG18 Agar plates are suitable for the enumeration of xerophilic molds in driedand semi-dried foods, such as dried fruits, meat and fish products, spices,confectionery, cereals, and nuts.

Legal Information

READYPLATE is a trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.462361.461891.46160
packaging

box of 20 plates

packaging

box of 20 or 120 plates

packaging

box of 20 plates

packaging

pkg of (90 mm plate (20 or per box))

plate diam.

90 mm

plate diam.

90 mm

plate diam.

55 mm

plate diam.

-

form

plate (ready-to-use)

form

plate (ready-to-use)

form

plate

form

plate (ready-to-use)

agency

ISO 21527

agency

EP, FDA-BAM, ISO, USP

agency

APHA, ISO 6888

agency

ISO 21871, ISO 7932

suitability

molds, yeasts

suitability

molds, yeasts

suitability

Staphylococcus spp., bacteria

suitability

Bacillus cereus

feature

ready-to-use

feature

ready-to-use

feature

-

feature

ready-to-use


Still not finding the right product?

Explore all of our products under DG 18 Agar


Storage Class

11 - Combustible Solids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters