Skip to Content
Merck
All Photos(2)

Key Documents

1.46137

Millipore

Baird-Parker Agar

ReadyPlate, ISO 6888, FDA-BAM, APHA, plate diam. 90 mm, gelatinous solid, box of 20 plates

Synonym(s):

Staphylococcus Selective Agar, Staphylococcus selective agar acc. to BAIRD-PARKER, Baird-Parker agar, ReadyPlate Baird Parker Agar ISO 6888, FDA-BAM

Sign Into View Organizational & Contract Pricing

Select a Size

20 PKG
CZK 1,060.00

CZK 1,060.00


Estimated to ship onApril 02, 2025Details


Request more information

Select a Size

Change View
20 PKG
CZK 1,060.00

About This Item

UNSPSC Code:
41106217
NACRES:
NA.74

CZK 1,060.00


Estimated to ship onApril 02, 2025Details


Request more information

Agency

APHA
FDA-BAM
ISO 6888

Quality Level

form

gelatinous solid

feature

ready-to-use

packaging

box of 20 plates

manufacturer/tradename

ReadyPlate

storage condition

protect from light (Store dry and tightly closed)

plate diam.

90 mm

color

yellow

pH

6.8 ( in H2O)

application(s)

environmental
food and beverages
microbiology

storage temp.

15-25°C

suitability

selective for Staphylococcus spp.

Looking for similar products? Visit Product Comparison Guide

General description

ReadyPlate Baird Parker Agar is used for the isolation and enumeration of Staphylococcus aureus. Potassium chloride, lithium chloride, and glycine allow the media to be selective for Staphylococcus aureus. Sodium pyruvate is essential to recover the damaged S. aureus cells and their subsequent growth.Agar is the solidifying agent. Staphylococcus colonies show two characteristic features when grown in this medium: (a) characteristic zones and rings are formed because of lipolysis and proteolysis, and (b) reduction of tellurite to tellurium produces a black coloration. The egg-yolk reaction and tellurite reduction usually occur together with a positive coagulase reaction and thereby serve as an index for the latter. According to EN ISO 6888-1, sulfamethazine addition is advised to suppress the growth and swarming of Proteus spp., if these are suspected in the test sample.

Application

ReadyPlate Baird Parker Agar is suitable for the isolation, presumptive identification, and enumeration of Staphylococcus aureus from food and animal feed, as well as other materials.

Legal Information

READYPLATE is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service