Skip to Content
Merck

Skip To

13-111

MBP, biotin conjugate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 mg

Estimated to ship onJune 25, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 13,900.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.41
eCl@ss:
32160405
Form:
liquid
Biological source:
bovine

CZK 13,900.00


Estimated to ship onJune 25, 2026Details


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine

Quality Segment

form

liquid

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable

UniProt accession no.

shipped in

wet ice

Application

Myelin basic protein (MBP) purified from bovine brain using SP-SepharoseTM high performance liquid chromatography and de-phosphorylated using Lambda protein phosphatase. Lambda protein phosphatase was inactivated, in the presence of 10mM sodium EDTA, at 65C for one hour. Purity was assessed at 95% using SDS-PAGE and Coomassie blue staining of 1μg of product. Developed for use in radioactive kinase assays.

Physical form

PBS, containing 0.05% sodium azide

Preparation Note

6 months at 4°C

Analysis Note

routinely evaluated for use as a substrate in a kinase assay

Other Notes

This product is derived from bovine source. Export of this product to certain countries may be restricted. Please contact Customer Service or your local distributor to inquire about product availability and export options.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
12-41412-38512-440
biological source

bovine

biological source

-

biological source

-

biological source

-

technique(s)

activity assay: suitable

technique(s)

activity assay: suitable (kinase)

technique(s)

activity assay: suitable (kinase)

technique(s)

activity assay: suitable

form

liquid

form

-

form

liquid

form

lyophilized

shipped in

wet ice

shipped in

dry ice

shipped in

dry ice

shipped in

wet ice

UniProt accession no.

P02686

UniProt accession no.

O14757

UniProt accession no.

-

UniProt accession no.

-

NCBI accession no.

NM_001025081.1, NM_001025092.1, NM_001025098.1, NM_001025101.1, NM_001025090.1, NM_001025100.1, NM_001025094.1, NM_002385.2

NCBI accession no.

NM_001274.3

NCBI accession no.

-

NCBI accession no.

-


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
13-11104053252371264
C4817-1EA04061826744123

Questions

Reviews

No rating value

Active Filters