Skip to Content
Merck
All Photos(1)

Key Documents

1.25079

Millipore

Fractogel® EMD DEAE (M)

MiniChrom Fractogel® DEAE (M), volume 5 mL, prepacked column

Synonym(s):

MiniChrom Column Fractogel® DEAE (M), 5 ml

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
Pricing and availability is not currently available.

ligand

diethyl-(2-hydroxypropyl)aminoethyl

material

polypropylene housing

product line

Fractogel®

form

prepacked column

parameter

≤8 bar pressure
~170 cm/hr max. flow rate

bed L

100 mm

bed volume

5 mL

column I.D.

8 mm

volume

5 mL

matrix active group

Methacrylate

mean particle size

40-90 μm

capacity

~100 mg, BSA binding capacity

application(s)

gene therapy
mAb
vaccine development

separation technique

anion exchange

storage temp.

15-25°C

General description

Weak cation exchange Fractogel® EMD DEAE (M) resin in a convenient pre-packed 5 ml column

Features and Benefits

MiniChrom Column Fractogel® DEAE (M), 5 ml enables:
  • Excellent binding to large viruses and plasmid DNA
  • Homogenous binding with high selectivity and purity
  • Lower elution volumes for the highest purity levels
  • Compatibility with 2.5 % (v/v) aqueous benzyl alcohol containing 150 mM NaCl storage solution


Due to the titration behavior, the ion exchange capacity can be used from pH 2 to pH 9.5. The separation of proteins is based on reversible electrostatic interactions between the negatively charged regions of the proteins′ surface and the support. Proteins are retained efficiently on Fractogel® EMD DEAE when the pH of the buffer is about 1 unit above their isoelectric points (pl).

The strength of the binding depends on the following:
  • the buffer system
  • pH value of the buffer which determines the surface charge of the protein
  • the degree of the ionization of the functional groups of the exchanger
  • the concentration of the counter ions
  • the charge density on the support (protein binding capacity)

Legal Information

FRACTOGEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service