Skip to Content
Merck

Skip To

120650

Eshmuno® CMX Resin

Eshmuno® CMX

Synonym(s):

Eshmuno® CMX

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mL
100 mL

Available to ship TODAYfromAreál Kühne+Nagel spol. s r.o.

CZK 22,500.00
500 mL

Available to ship TODAYfromAreál Kühne+Nagel spol. s r.o.

CZK 112,400.00
5 L

About This Item

UNSPSC Code:
47101511

This item is not able to be purchased as a guest user. For further information, please contact us at +420 225 296 100or

.

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


ligand

(Carboxy and Alkyl Group)

Quality Segment

sterility

sterile (Caustic Stable)

form

resin

specific analyte(s)

MAbs

parameter

300 cm/hr flow rate, 8 bar max. pressure

matrix active group

polymer, polyvinyl ether

mean particle size

50 μm

capacity

60 mg binding capacity (pIgG/ml of resin)

application(s)

ADC
mAb

separation technique

mixed mode

storage temp.

2-30°C

General description

Eshmuno® CMX chromatography resin is a mixed mode chromatography resin built on the proven Eshmuno® resin technology. This innovative resin combines weak cation exchange properties with hydrophobic interaction, providing high selectivity for Monoclonal Antibody (mAb), fusion protein and Antibody Drug Conjugate (ADCs) purification as well as separation of low molecular weight impurities, Host Cell Proteins (HCPs) and glycovariants.

Application

Purification of mAbs, ADCs, fusion proteins

Features and Benefits

Eshmuno® CMX chromatography resin enables users to
  • Intensify processes and decrease costs by reducing the number of chromatographic steps and buffer consumption
  • Improve performance with higher recovery rates, high selectivity and superior dynamic binding capacity
  • Improve the user experience with a broad operational window, simplified process development and easy column packing due to a rigid base bead

Legal Information

Eshmuno is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
125187M125188M125185M
ligand

(Carboxy and Alkyl Group)

ligand

(Carboxyethyl with Alkyl-Functional Groups)

ligand

(Carboxyethyl with Alkyl-Functional Groups)

ligand

(Carboxyethyl with Alkyl-Functional Groups)

form

resin

form

prepacked column

form

prepacked column

form

prepacked column

separation technique

mixed mode

separation technique

-

separation technique

-

separation technique

-

capacity

60 mg binding capacity (pIgG/ml of resin)

capacity

~60 mL, resin binding capacity

capacity

~60 mL, resin binding capacity

capacity

~60 mL, resin binding capacity

matrix active group

polymer, polyvinyl ether

matrix active group

Polyvinyl Ether

matrix active group

Polyvinyl Ether

matrix active group

Polyvinyl Ether

specific analyte(s)

MAbs

specific analyte(s)

-

specific analyte(s)

-

specific analyte(s)

-


Still not finding the right product?

Explore all of our products under Eshmuno® CMX Resin


pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk

WGK 3

flash_point_f

95.0 °F

flash_point_c

35 °C



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters