Skip to Content
Merck

Skip To

12-459

IKK Substrate Peptide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 mg

Estimated to ship onMay 04, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 7,810.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.42
eCl@ss:
32160405

CZK 7,810.00


Estimated to ship onMay 04, 2026Details


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

Upstate®

Quality Level

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... CHUK(1147)

Biochem/physiol Actions

Protein Target: IKKα
Target Sub-Family: Misc

Physical form

Lyophilized powder

Preparation Note

Lyophilized: 2 years at 4°C; Rehydrated: 6 months at -20°C

Analysis Note

Routinely evaluated as a substrate for IKKα (Catalog #14-461) and IKKβ (Catalog #14-485)

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
12-12312-34012-152
technique(s)

activity assay: suitable (kinase)

technique(s)

activity assay: suitable (kinase)

technique(s)

activity assay: suitable (kinase)

technique(s)

activity assay: suitable (kinase)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

Gene Information

human ... CHUK(1147)

Gene Information

human ... PRKCA(5578)

Gene Information

-

Gene Information

-

UniProt accession no.

O15111

UniProt accession no.

P17252

UniProt accession no.

P31749

UniProt accession no.

P17612

shipped in

dry ice

shipped in

wet ice

shipped in

dry ice

shipped in

wet ice

manufacturer/tradename

Upstate®

manufacturer/tradename

Upstate®

manufacturer/tradename

Upstate®

manufacturer/tradename

Upstate®


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
12-45904053252736889

Questions

Reviews

No rating value

Active Filters