Skip to Content
Merck

Skip To

12-427

Phospho-Histone H3 Peptide

lyophilized powder, biotin conjugate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μg

Estimated to ship onMay 27, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 9,660.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

CZK 9,660.00


Estimated to ship onMay 27, 2026Details


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Phospho (Ser10)-Histone H3 Peptide, biotin conjugate, Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes.

Quality Level

form

lyophilized powder

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... H3F3B(3021)

Application

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes.

Physical form

Lyophilized powder

Preparation Note

2 years at -20°C

Analysis Note

Routinely evaluated as a substrate for histone-modifying enzymes.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
12-42812-40312-569
technique(s)

immunoprecipitation (IP): suitable

technique(s)

immunoprecipitation (IP): suitable

technique(s)

immunoprecipitation (IP): suitable

technique(s)

immunoprecipitation (IP): suitable

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

form

lyophilized powder

form

-

form

-

form

-

UniProt accession no.

Q16695

UniProt accession no.

Q16695

UniProt accession no.

Q16695

UniProt accession no.

Q16695

shipped in

dry ice

shipped in

dry ice

shipped in

wet ice

shipped in

dry ice

Gene Information

human ... H3F3B(3021)

Gene Information

human ... H3F3B(3021)

Gene Information

human ... H3F3B(3021)

Gene Information

human ... H3F3B(3021)


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
12-42704053252508899

Questions

Reviews

No rating value

Active Filters