Skip to Content
Merck

Skip To

1.19780

Chromate standard solution

traceable to SRM of NIST K2CrO4 in H2O 1000 mg/l CrO4 Certipur®

Synonym(s):

Chromate standard for aqueous matrices ready foruse, Nota 1 A

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107


Quality Segment

technique(s)

ion chromatography: suitable

pH

7 (20 °C in H2O)

density

1.0 g/cm3 at 20 °C

application(s)

environmental
food and beverages

format

single component solution

storage temp.

15-25°C

Analysis Note

Concentration     β (CrO₄²⁻): 990 - 1010 mg/l
Determination method: Iodometric titration.
The content of this solution was determined with sodium thiosulfate standard solution (article number 1.09147) standardized against volumetric standard potassium iodate (article number 1.02404). The expanded measurement uncertainty is ± 0.4 % (k=2 coverage factor for 95% coverage probability). The certified value is traceable to primary standard NIST SRM 136f (NIST: National Institute of Standards and Technology, USA) by means of volumetric standard potassium iodate, measured in the accredited calibration laboratory of Merck KGaA, Darmstadt, Germany in accordance to DIN EN ISO/IEC 17025.

Other Notes

We now offer a certified reference material alternative for this product that has the same composition and is manufactured under ISO 17034 accreditation.

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.198961.197961.19785
technique(s)

ion chromatography: suitable

technique(s)

ion chromatography: suitable

technique(s)

AAS: suitable

technique(s)

AAS: suitable

application(s)

environmental
food and beverages

application(s)

environmental
food and beverages

application(s)

industrial qc
pharmaceutical

application(s)

industrial qc
pharmaceutical

format

single component solution

format

single component solution

format

single component solution

format

single component solution

pH

7 (20 °C in H2O)

pH

-

pH

0.5 (20 °C in H2O)

pH

0.5 (20 °C in H2O)

density

1.0 g/cm3 at 20 °C

density

0.999 g/cm3 at 20 °C

density

1.013 g/cm3 at 20 °C

density

1.014 g/cm3 at 20 °C

storage temp.

15-25°C

storage temp.

15-25°C

storage temp.

15-25°C

storage temp.

15-25°C


Still not finding the right product?

Explore all of our products under Chromate standard solution


pictograms

Health hazard

signalword

Danger

Hazard Classifications

Carc. 1B Inhalation - Muta. 1B

Storage Class

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

7789-00-6

CAS No.

7789-00-6

CAS No.

7789-00-6

CAS No.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Undisputed Accuracy for Your Quantitative ResultsDiluted ready-to-use andCertified Reference Materials forwater and wastewater analysis

View All Related Content




Global Trade Item Number

SKUGTIN
119780050004022536180540

Questions

Reviews

No rating value

Active Filters