Skip to Content
Merck

Skip To

1.19157

Standard samples for HPLC system qualification

suitable for HPLC, methyl paraben in methanol/water 50-200 mg/L, for use with LICHROTEST

Synonym(s):

TM=["LICHROTEST"] Standard samples for HPLC system qualification Set 1 A Precision and Linearity

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 units
Please contact Customer Service for Availability
CZK 5,160.00

About This Item

NACRES:
NA.24
UNSPSC Code:
41115709

CZK 5,160.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

composition

methyl paraben in methanol/water, 50-200 mg/L

technique(s)

HPLC: suitable

compatibility

for use with LICHROTEST

format

mixture

storage temp.

15-25°C

General description

Set 1 A: Precision and Linearity; Parabens in methanol/water (50:50) LiChroTest


In our effort to be more sustainable, we′re going digital with COAs-download them directly from our website. Download your certificate at https://www.sigmaaldrich.com/US/en/documents-search to view certified values, including uncertainty and expiry date.

Analysis Note

50 mg/l Methyl paraben: in accordance
100 mg/l Methyl paraben: in accordance
150 mg/l Methyl paraben: in accordance
200 mg/l Methyl paraben: in accordance
Linearity (> 0.9999): in accordance

Legal Information

LICHROTEST is a trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.191581.191611.19162
format

mixture

format

mixture

format

mixture

format

mixture

technique(s)

HPLC: suitable

technique(s)

HPLC: suitable

technique(s)

HPLC: suitable

technique(s)

HPLC: suitable

grade

analytical standard

grade

analytical standard

grade

analytical standard

grade

analytical standard

composition

methyl paraben in methanol/water, 50-200 mg/L

composition

methyl paraben in methanol/water, 100 mg/L

composition

methyl paraben in methanol, 60 mg/L

composition

methyl paraben in methanol, 1.5-150 mg/L

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

compatibility

for use with LICHROTEST

compatibility

for use with LICHROTEST

compatibility

for use with LICHROTEST

compatibility

for use with LICHROTEST


Still not finding the right product?


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class

3 - Flammable liquids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters