Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
300 tests | Please contact Customer Service for Availability | CZK 4,360.00 |
About This Item
CZK 4,360.00
Product Name
Chromate Test, colorimetric, 0.10-1.6 mg/L (Cr), 0.22-3.6 mg/L (CrO42-), for use with MCOLORTEST®
specific analyte(s)
chromate
Quality Segment
measuring range
0.10-1.6 mg/L (Cr), 0.22-3.6 mg/L (CrO42-)
compatibility
for use with MCOLORTEST®
detection method
colorimetric
storage temp.
15-25°C
General description
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| detection method colorimetric | detection method colorimetric | detection method colorimetric | detection method photometric (Diphenylcarbazide) |
| specific analyte(s) chromate | specific analyte(s) chromate | specific analyte(s) chromate | specific analyte(s) chromate |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| measuring range 0.10-1.6 mg/L (Cr), 0.22-3.6 mg/L (CrO42-) | measuring range 0.005-0.10 mg/L (Cr), 0.011-0.22 mg/L (CrO42-) | measuring range 0.10-10 mg/L (Cr), 0.22-22 mg/L (CrO42-) | measuring range 0.010-3.00 mg/L (Cr), 0.02-6.69 mg/L (CrO4) |
| compatibility for use with MCOLORTEST® | compatibility for use with MQUANT® | compatibility for use with MQUANT® | compatibility for use with Spectroquant® Move 100, for use with Spectroquant® Nova 60 A, for use with Spectroquant® Prove 100 (plus), for use with Spectroquant® Prove 300 (plus), for use with Spectroquant® Prove 600 (plus) |
| storage temp. 15-25°C | storage temp. 15-25°C | storage temp. 15-25°C | storage temp. 15-25°C |
Still not finding the right product?
Explore all of our products under Chromate Test
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2
Storage Class
8A - Combustible, corrosive hazardous materials
wgk
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



