Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
200 tests | Please contact Customer Service for Availability | CZK 3,220.00 |
About This Item
CZK 3,220.00
Product Name
Nitrate Test, colorimetric, 10-150 mg/L (NO3-), for use with MQUANT®
specific analyte(s)
nitrate
Quality Segment
measuring range
10-150 mg/L (NO3-)
compatibility
for use with MQUANT®
detection method
colorimetric
storage temp.
15-25°C
General description
- Groundwater, drinking water, and surface water (also humin-containing samples)
- Waters from aquaculture
- Swimming-pool water
- Industrial water
- Wastewater
- Food and animal fodder after appropriate sample pretreatment
- Soils and fertilizers after appropriate sample pretreatment
Method
Nitrate ions are reduced to nitrite ions that react in acidic solution with sulfanilic acid to form a diazonium salt. This reacts with a benzoic acid derivative to form an orange-yellow azo dye. The nitrate concentration is measured semiquantitatively by allocating the color to a value on a reference scale.
Analysis Note
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| specific analyte(s) nitrate | specific analyte(s) nitrate | specific analyte(s) nitrate | specific analyte(s) nitrate |
| detection method colorimetric | detection method colorimetric | detection method colorimetric | detection method colorimetric |
| compatibility for use with MQUANT® | compatibility for use with MQUANT® | compatibility - | compatibility for use with MQuant® StripScan App |
| storage temp. 15-25°C | storage temp. 15-25°C | storage temp. 2-8°C | storage temp. 15-25°C |
| measuring range 10-150 mg/L (NO3-) | measuring range 5-90 mg/L (NO3-) | measuring range 10-500 mg/L (NO3-) | measuring range 10-500 mg/L (NO3-) |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
Still not finding the right product?
Explore all of our products under Nitrate Test
signalword
Danger
Hazard Classifications
Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Repr. 1B - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Inhalation
Storage Class
6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.






