Skip to Content
Merck

Skip To

1.11131

Magnesium Test

colorimetric, 100-1500 mg/L (Mg), for use with MCOLORTEST®

Synonym(s):

Magnesium Visual Test Kit

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
41116116


specific analyte(s)

magnesium

Quality Segment

measuring range

100-1500 mg/L (Mg)

compatibility

for use with MCOLORTEST®

detection method

colorimetric

storage temp.

15-25°C

General description

Method: colorimetric with color card 100 - 200 - 300 - 500 - 1000 - 1500 mg/l Mg MQuant®
This MQuant® Magnesium test (formally MColortest®) is suitable for the colorimetric determination using the easy to use color card.
It can be used for samples like
  • Groundwater and surface water
  • Drinking water

Method
Magnesium ions react with xylidyl blue (Mann and Yoe′s reagent) to form a red dye. The magnesium concentration is measured semiquantitatively by visual comparison of the color of the measurement solution with the color fields of a color card.

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.144021.161241.14660
specific analyte(s)

magnesium

specific analyte(s)

chromate

specific analyte(s)

magnesium

specific analyte(s)

iron

detection method

colorimetric

detection method

colorimetric

detection method

reflectometric

detection method

colorimetric

storage temp.

15-25°C

storage temp.

15-25°C

storage temp.

2-8°C

storage temp.

15-25°C

compatibility

for use with MCOLORTEST®

compatibility

for use with MQUANT®

compatibility

RQflex® 20, for use with REFLECTOQUANT®

compatibility

for use with MQUANT®

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

measuring range

100-1500 mg/L (Mg)

measuring range

0.005-0.10 mg/L (Cr), 0.011-0.22 mg/L (CrO42-)

measuring range

5-100 mg/L (Mg)

measuring range

0.05-1.0 mg/L (Fe2+)


signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Repr. 1B

Storage Class

3 - Flammable liquids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters