Skip to Content
Merck
All Photos(1)

Key Documents

1.10823

Millipore

L-PALCAM-Listeria selective enrichment broth (Base) acc. to van Netten et al.

for microbiology

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
CZK 4,210.00

CZK 4,210.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
500 G
CZK 4,210.00

About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

CZK 4,210.00


Please contact Customer Service for Availability

Request more information

Quality Level

form

powder (dehydrated (DCM))

manufacturer/tradename

GranuCult®

technique(s)

microbiological culture: suitable

pH

7.3-7.5 (25 °C, 47.33 g/L in H2O)

solubility

47.33 g/L

bulk density

700 kg/m3

application(s)

food and beverages
microbiology
pathogen testing

storage temp.

15-25°C

suitability

Listeria spp.

Analysis Note

Base tested with PALCAM-Listeria selective supplement acc. to VAN NETTEN et.al. art. no. 1.12122.
Appearance (clearness): opalescent to turbid
Appearance (colour): red
pH-value (25 °C): 7.2 - 7.6
Growth (Listeria monocytogenes ATCC 35152 (WDCM 00109)): good
Colour change to brown-black (Listeria monocytogenes ATCC 35152 (WDCM 00109)): +
Growth (Listeria monocytogenes ATCC 19114 (WDCM 00110)): good
Colour change to brown-black (Listeria monocytogenes ATCC 19114 (WDCM 00110)): +
Growth (Listeria monocytogenes ATCC 13932 (WDCM 00021)): good
Colour change to brown-black (Listeria monocytogenes ATCC 13932 (WDCM 00021)): +
Growth (Listeria ivanovii ATCC 19119 (WDCM 00018)): good
Colour change to brown-black (Listeria ivanovii ATCC 19119 (WDCM 00018)): +
Growth (Enterococcus faecium ATCC 882): inhibited
Growth (Kocuria rhizophila ATCC 9341): inhibited
Growth (Staphylococcus aureus ATCC 6538 (WDCM 00032)): inhibited
Incubation: 24 - 48 hrs.; 30 °C; aerobic.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Detection of Listeria monocytogenes in food and environmental samples according to ISO 11290-1, FDA-BAM and USDA-FSIS

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service