Skip to Content
Merck

Skip To

1.09872

Sulfate standard

1000 mg SO4 (H2SO4 in H2O) Titrisol®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 amp
Please contact Customer Service for Availability
CZK 1,050.00

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107

CZK 1,050.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Titrisol®

technique(s)

ion chromatography: suitable

pH

0.8 (20 °C in H2O)

density

1.01 g/cm3 at 20 °C

application(s)

environmental
food and beverages

format

single component solution

storage temp.

15-25°C

Analysis Note

content (SO₄²⁻): 1000 mg / Ampoule
Determination method: Alcalimetric titration
(traceable to NIST - SRM 723b)
Accuracy of the method: +/- 2 mg

Legal Information

Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.250531.250511.25052
application(s)

environmental
food and beverages

application(s)

agriculture
biotechnology/fermenter
construction materials
disposal drainage water
drinking water
environmental
ground and surface water
mineral water
mining
power plants
seawater
wastewater

application(s)

agriculture
biotechnology/fermenter
construction materials
disposal drainage water
drinking water
environmental
ground and surface water
mineral water
mining
power plants
seawater
wastewater

application(s)

agriculture
biotechnology/fermenter
construction materials
disposal drainage water
drinking water
environmental
ground and surface water
mineral water
mining
power plants
seawater
wastewater

technique(s)

ion chromatography: suitable

technique(s)

photometry: suitable

technique(s)

photometry: suitable

technique(s)

photometry: suitable

format

single component solution

format

single component solution

format

single component solution

format

single component solution

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

200

storage temp.

15-25°C

storage temp.

15-25°C

storage temp.

15-25°C

storage temp.

15-25°C

density

1.01 g/cm3 at 20 °C

density

-

density

-

density

-


pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

8B - Non-combustible, corrosive hazardous materials

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters