Skip to Content
Merck

Skip To

1.08208

Triethylammonium acetate buffer solution

for the purification of chemically synthesized oligonucleotides (1 mol/l) pH 7,0

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.05
UNSPSC Code:
12161700
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

23 hPa ( 20 °C)

Quality Segment

form

liquid

pH

7.0 (20 °C in H2O)

bp

100 °C

density

1 g/cm3 at 20 °C

storage temp.

15-25°C

General description

for the purification of chemically synthesized oligonucleotides (1 mol/l) pH 7,0

Analysis Note

Evaporation residue≤ 0.01 %pH-value (1 mol/l)6.9 - 7.1


Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content






Global Trade Item Number

SKUGTIN
108208100004022536101675

Questions

Reviews

No rating value

Active Filters