Skip to Content
Merck

Skip To

1.08181

Thiamine chloride hydrochloride

for biochemistry

Synonym(s):

Thiamine chloride hydrochloride, Aneurin hydrochloride, Vitamin B1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 g
Please contact Customer Service for Availability
CZK 1,340.00
250 g
Please contact Customer Service for Availability
CZK 6,890.00

About This Item

Empirical Formula (Hill Notation):
C12H17ClN4OS · xHCl
CAS Number:
Molecular Weight:
300.81 (free base basis)
UNSPSC Code:
12352201
EC Index Number:
200-641-8
NACRES:
NA.21
MDL number:

CZK 1,340.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

solid

potency

3710 mg/kg LD50, oral (Rat)

pH

2.7-3.3 (20 °C, 25 g/L in H2O)

mp

248 °C (decomposition)

bulk density

500 kg/m3

storage temp.

15-25°C

SMILES string

[s]1c[n+](c(c1CCO)C)Cc2c(nc(nc2)C)N.[Cl-].[Cl-].[H+]

InChI

1S/C12H17N4OS.2ClH/c1-8-11(3-4-17)18-7-16(8)6-10-5-14-9(2)15-12(10)13;;/h5,7,17H,3-4,6H2,1-2H3,(H2,13,14,15);2*1H/q+1;;/p-1

InChI key

DPJRMOMPQZCRJU-UHFFFAOYSA-M

Analysis Note

Identity (IR-spectrum): conforms
Assay (Perchloric acid titration, calc. on anhydrous substance): ≥ 99.0 %
Sulphate (SO₄): ≤ 0.01 %
Nitrate (NO₃): ≤ 0.005 %
Heavy metals (as Pb): ≤ 0.001 %
Water: ≤ 5.0 %

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1656002Y00004675871
form

solid

form

-

form

-

form

solid

storage temp.

15-25°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

15-25°C

Quality Level

100

Quality Level

-

Quality Level

-

Quality Level

100

mp

248 °C (decomposition)

mp

250 °C (dec.) (lit.)

mp

250 °C (dec.) (lit.)

mp

-

potency

3710 mg/kg LD50, oral (Rat)

potency

-

potency

-

potency

-

bulk density

500 kg/m3

bulk density

-

bulk density

-

bulk density

-


Still not finding the right product?

Explore all of our products under Thiamine chloride hydrochloride


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters