Skip to Content
Merck

Skip To

1.07769

L-Serine

for biochemistry

Synonym(s):

L-Serine, 3-Hydroxy-L-alanine, Ser

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 g
Please contact Customer Service for Availability
CZK 1,400.00
100 g
Please contact Customer Service for Availability
CZK 3,250.00
1 kg
Please contact Customer Service for Availability
CZK 24,700.00
10 kg
Please contact Customer Service for Availability
CZK 119,200.00

About This Item

Empirical Formula (Hill Notation):
C3H7NO3
CAS Number:
Molecular Weight:
105.09
UNSPSC Code:
12352209
EC Index Number:
200-274-3
NACRES:
NA.21
MDL number:
eCl@ss:
32160406

CZK 1,400.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

solid

pH

5-6 (20 °C, 100 g/L in H2O)

mp

215-225 °C (decomposes)

solubility

360 g/L

bulk density

570 kg/m3

storage temp.

2-30°C

SMILES string

[N+H3]C(CO)C(=O)[O-]

InChI

1S/C3H7NO3/c4-2(1-5)3(6)7/h2,5H,1,4H2,(H,6,7)

InChI key

MTCFGRXMJLQNBG-UHFFFAOYSA-N

Analysis Note

Assay (perchloric acid titration, calculated on dried substance): ≥ 99.0 %
Identity (IR-spectrum): passes test
Appearance: white to almost white or colorless, needle-shaped crystals
Appearance of solution (1 mol/l, water): clear and colorless
Spec. rotation (α 20/D, 50 g/l, hydrochloric acid 1 mol/l, calc. on dried substance): +14.3 to +15.8
UV-Absorption(260 nm; 1 mol/l; 1 cm; water): ≤ 0.05
UV-Absorption(280 nm; 1 mol/l; 1 cm; water): ≤ 0.02
Chloride (Cl): ≤ 0.02 %
Sulfate (SO₄): ≤ 0.01 %
Heavy metals (as Pb): ≤ 0.001 %
As (Arsenic): ≤ 0.0005 %
Ca (Calcium): ≤ 0.001 %
Co (Cobalt): ≤ 0.0005 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.0005 %
Mg (Magnesium): ≤ 0.0005 %
Na (Sodium): ≤ 0.005 %
NH₄ (Ammonium): ≤ 0.01 %
Zn (Zinc): ≤ 0.0005 %
Foreign amino acids: ≤ 0.3 %
Other ninhydrine positive substances: ≤ 0.1 %
Loss on drying (105 °C; 3 h): ≤ 0.3 %

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.076478.16020S0450000
Serine European Pharmacopoeia (EP) Reference Standard

S0450000

Serine

form

solid

form

crystalline powder

form

solid

form

-

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-8°C

solubility

360 g/L

solubility

360 g/L

solubility

360 g/L

solubility

-

mp

215-225 °C (decomposes)

mp

215-225 °C (decomposes)

mp

215-225 °C (decomposes)

mp

222 °C (dec.) (lit.)

Quality Level

100

Quality Level

500

Quality Level

200

Quality Level

-

pH

5-6 (20 °C, 100 g/L in H2O)

pH

5-6 (20 °C, 100 g/L in H2O)

pH

5-6 (20 °C, 100 g/L in H2O)

pH

-


Still not finding the right product?

Explore all of our products under L-Serine


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters