Skip to Content
Merck

Skip To

1.07433

Pronase E

Synonym(s):

Streptomyces grisens neutral proteinase

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.21
UNSPSC Code:
12352204


form

solid

potency

3290 mg/kg LD50, oral (rat)

pH

6-7 (20 °C, 10 g/L in H2O)

storage temp.

2-8°C

General description

Pronase E is a mixture of proteolytic enzymes that is obtained from Streptomyces griseus. It can be used in the in diagnostics in histopathology as it hydrolyses proteins into individual amino acids.
from Streptomyces griseus 4,000,000 PU/g for biochemistry EC 3.4.24.4

Application

Pronase E (from Streptomyces griseus) 4,000,000 PU/g for biochemistry EC 3.4.24.4. CAS 9036-06-0, pH 6 - 7 (10 g/l, H₂O, 20 °C).

Analysis Note

Appearance (colour):slightly brown
Appearance (description):fine powder
Activity (Casein; pH 7.4; 40 °C):≥ 4000000 U/g

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
7433P8811P5147
Pronase E

7433

Pronase E

form

solid

form

fine powder

form

powder

form

powder

pH

6-7 (20 °C, 10 g/L in H2O)

pH

-

pH

-

pH

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

potency

3290 mg/kg LD50, oral (rat)

potency

-

potency

-

potency

-


pictograms

Exclamation markHealth hazard

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters